Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
Chromosome Location
Chr.12: 121402289 - 121582268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032590.4 NP_115979.3 16 - 17 2559 75
XM_005253955.3 XP_005254012.1 16 - 17 2769 75
XM_005253956.3 XP_005254013.1 16 - 17 2366 75
XM_005253961.4 XP_005254018.1 5 - 6 854 75
XM_011538867.2 XP_011537169.1 16 - 17 3821 75
XM_011538868.2 XP_011537170.1 16 - 17 3817 75
XM_011538869.2 XP_011537171.1 16 - 17 3819 75
XM_011538875.2 XP_011537177.1 5 - 6 863 75
GenBank mRNA AB031230.1 - 6 - 7 1027 75
AJ459424.1 - 16 - 17 2531 75
AK027440.1 - 6 - 7 948 75
AK027692.1 - 3 - 4 464 75
AK074718.1 - 1 - 2 43 75
AK299827.1 - 5 - 6 873 75
AK316470.1 - 5 - 6 866 75
AL133572.1 - 11 - 12 2810 75
BC008735.2 - 2 - 3 329 75

View Full Product Details