Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TANGO2
- Entrez Gene ID
-
128989
- Gene Name
- transport and golgi organization 2 homolog
- Gene Aliases
-
C22orf25, MECRCN
- Chromosome Location
-
Chr.22: 20017000 - 20067164 on Build GRCh38
View Full Product Details