Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41866515-41867056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACLY
Entrez Gene ID
47
Gene Name
ATP citrate lyase
Gene Aliases
ACL, ATPCL, CLATP
UniGene
Hs.387567
Gene Chromosome Location
Chr.17: 41866916-41930542 on Build GRCh38
Transcripts
NM_001096.2 NM_001303274.1 NM_001303275.1 NM_198830.1 XM_005257395.1 XM_017024688.1

SNP Detail

SNP ID Chromosome Location
rs782275979 Chr.17: 41866948-41866948 on Build GRCh38
rs66811705 Chr.17: 41866656-41866658 on Build GRCh38
rs956701992 Chr.17: 41866883-41866883 on Build GRCh38
rs782430488 Chr.17: 41866714-41866714 on Build GRCh38
rs9303321 Chr.17: 41866657-41866657 on Build GRCh38
rs558442312 Chr.17: 41867019-41867019 on Build GRCh38
rs1058928 Chr.17: 41867005-41867005 on Build GRCh38
rs1030150877 Chr.17: 41866909-41866909 on Build GRCh38
rs942046147 Chr.17: 41866577-41866577 on Build GRCh38
rs904744246 Chr.17: 41866742-41866742 on Build GRCh38
rs112685370 Chr.17: 41866869-41866869 on Build GRCh38
rs9303319 Chr.17: 41866556-41866556 on Build GRCh38
rs972510904 Chr.17: 41866954-41866954 on Build GRCh38
rs934647 Chr.17: 41866772-41866772 on Build GRCh38
rs386797131 Chr.17: 41866676-41866679 on Build GRCh38
rs953199304 Chr.17: 41866920-41866920 on Build GRCh38
rs191273121 Chr.17: 41866660-41866660 on Build GRCh38
rs547037791 Chr.17: 41866726-41866726 on Build GRCh38
rs938791823 Chr.17: 41867004-41867004 on Build GRCh38
rs938774632 Chr.17: 41866634-41866634 on Build GRCh38
rs9303322 Chr.17: 41866676-41866676 on Build GRCh38
rs1036339637 Chr.17: 41866579-41866579 on Build GRCh38
rs9303320 Chr.17: 41866656-41866656 on Build GRCh38
rs915972870 Chr.17: 41867023-41867023 on Build GRCh38
rs997267709 Chr.17: 41866886-41866886 on Build GRCh38
rs985998212 Chr.17: 41866547-41866547 on Build GRCh38
rs545971997 Chr.17: 41866675-41866675 on Build GRCh38
rs894632512 Chr.17: 41866710-41866710 on Build GRCh38
rs963293777 Chr.17: 41866950-41866950 on Build GRCh38
rs919345744 Chr.17: 41866588-41866588 on Build GRCh38
rs111326058 Chr.17: 41866731-41866731 on Build GRCh38
rs569956064 Chr.17: 41866951-41866951 on Build GRCh38
rs111597057 Chr.17: 41866677-41866677 on Build GRCh38
rs9944425 Chr.17: 41866678-41866678 on Build GRCh38
rs563786739 Chr.17: 41866975-41866975 on Build GRCh38
rs574337186 Chr.17: 41866616-41866616 on Build GRCh38
rs201686569 Chr.17: 41866654-41866658 on Build GRCh38
rs985904387 Chr.17: 41866926-41866926 on Build GRCh38
rs199763985 Chr.17: 41866655-41866656 on Build GRCh38

View Full Product Details