Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41873728-41874199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACLY
Entrez Gene ID
47
Gene Name
ATP citrate lyase
Gene Aliases
ACL, ATPCL, CLATP
UniGene
Hs.387567
Gene Chromosome Location
Chr.17: 41866916-41930542 on Build GRCh38
Transcripts
NM_001096.2 NM_001303274.1 NM_001303275.1 NM_198830.1 XM_005257395.1 XM_017024688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644250 c.2637G>C p.Q879H Chr.17: 41873816-41873816 on Build GRCh38
3517581 c.2623C>T p.L875F Chr.17: 41873830-41873830 on Build GRCh38
1644251 c.2499G>C p.L833F Chr.17: 41873954-41873954 on Build GRCh38
3728704 c.2610C>T p.G870G Chr.17: 41873843-41873843 on Build GRCh38
6376598 c.2572C>G p.P858A Chr.17: 41873881-41873881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782321787 Chr.17: 41873840-41873840 on Build GRCh38
rs115627685 Chr.17: 41873761-41873761 on Build GRCh38
rs2277697 Chr.17: 41873834-41873834 on Build GRCh38
rs781897910 Chr.17: 41873980-41873980 on Build GRCh38
rs781955124 Chr.17: 41873873-41873873 on Build GRCh38
rs782554718 Chr.17: 41873764-41873764 on Build GRCh38
rs979196894 Chr.17: 41873804-41873804 on Build GRCh38
rs1012304344 Chr.17: 41874107-41874107 on Build GRCh38
rs1000689594 Chr.17: 41874139-41874139 on Build GRCh38
rs781806692 Chr.17: 41873770-41873770 on Build GRCh38
rs376696137 Chr.17: 41874010-41874010 on Build GRCh38
rs967928056 Chr.17: 41873803-41873803 on Build GRCh38
rs151017433 Chr.17: 41874142-41874142 on Build GRCh38
rs1030632789 Chr.17: 41874170-41874170 on Build GRCh38
rs976094580 Chr.17: 41873887-41873887 on Build GRCh38
rs536480528 Chr.17: 41874169-41874169 on Build GRCh38
rs782488395 Chr.17: 41874013-41874013 on Build GRCh38
rs542002258 Chr.17: 41874073-41874073 on Build GRCh38
rs781846133 Chr.17: 41873942-41873942 on Build GRCh38
rs782489504 Chr.17: 41873797-41873797 on Build GRCh38
rs565966334 Chr.17: 41874164-41874164 on Build GRCh38
rs782797920 Chr.17: 41873971-41873971 on Build GRCh38
rs781894956 Chr.17: 41873762-41873762 on Build GRCh38
rs186187040 Chr.17: 41873808-41873808 on Build GRCh38
rs370576290 Chr.17: 41874046-41874046 on Build GRCh38
rs530924991 Chr.17: 41874089-41874089 on Build GRCh38
rs139677928 Chr.17: 41874117-41874117 on Build GRCh38
rs142674352 Chr.17: 41874140-41874140 on Build GRCh38
rs953423851 Chr.17: 41874177-41874177 on Build GRCh38
rs898111945 Chr.17: 41874047-41874047 on Build GRCh38
rs782501478 Chr.17: 41873968-41873968 on Build GRCh38
rs782247733 Chr.17: 41873806-41873806 on Build GRCh38
rs140430864 Chr.17: 41873888-41873888 on Build GRCh38
rs559615450 Chr.17: 41873991-41873991 on Build GRCh38
rs781924155 Chr.17: 41873918-41873918 on Build GRCh38
rs375322610 Chr.17: 41873912-41873912 on Build GRCh38
rs896262307 Chr.17: 41873755-41873755 on Build GRCh38
rs117473633 Chr.17: 41873936-41873936 on Build GRCh38
rs782616451 Chr.17: 41873799-41873799 on Build GRCh38
rs547756441 Chr.17: 41874152-41874152 on Build GRCh38
rs782624246 Chr.17: 41874016-41874016 on Build GRCh38
rs782729574 Chr.17: 41873937-41873937 on Build GRCh38
rs151013779 Chr.17: 41873817-41873817 on Build GRCh38
rs782813132 Chr.17: 41873897-41873897 on Build GRCh38
rs782209089 Chr.17: 41874012-41874012 on Build GRCh38
rs782702318 Chr.17: 41873768-41873768 on Build GRCh38
rs782376333 Chr.17: 41873886-41873886 on Build GRCh38
rs938910731 Chr.17: 41874066-41874066 on Build GRCh38
rs782224299 Chr.17: 41873878-41873878 on Build GRCh38
rs903568267 Chr.17: 41874118-41874118 on Build GRCh38
rs563512930 Chr.17: 41874074-41874074 on Build GRCh38
rs782154200 Chr.17: 41873891-41873891 on Build GRCh38
rs781987769 Chr.17: 41873975-41873975 on Build GRCh38
rs782793636 Chr.17: 41873994-41873994 on Build GRCh38

View Full Product Details