Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41878562-41878989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACLY
Entrez Gene ID
47
Gene Name
ATP citrate lyase
Gene Aliases
ACL, ATPCL, CLATP
UniGene
Hs.387567
Gene Chromosome Location
Chr.17: 41866916-41930542 on Build GRCh38
Transcripts
NM_001096.2 NM_001303274.1 NM_001303275.1 NM_198830.1 XM_005257395.1 XM_017024688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424627 c.2393+7G>A p.? Chr.17: 41878790-41878790 on Build GRCh38
7180979 c.2337G>A p.L779L Chr.17: 41878853-41878853 on Build GRCh38
5386370 c.2269C>T p.Q757* Chr.17: 41878921-41878921 on Build GRCh38
3188014 c.2350G>T p.V784L Chr.17: 41878840-41878840 on Build GRCh38
4480141 c.2391C>T p.I797I Chr.17: 41878799-41878799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913222298 Chr.17: 41878600-41878600 on Build GRCh38
rs782227179 Chr.17: 41878960-41878960 on Build GRCh38
rs532511120 Chr.17: 41878861-41878861 on Build GRCh38
rs561409529 Chr.17: 41878593-41878593 on Build GRCh38
rs782635277 Chr.17: 41878888-41878888 on Build GRCh38
rs782181054 Chr.17: 41878931-41878931 on Build GRCh38
rs371442891 Chr.17: 41878945-41878945 on Build GRCh38
rs781968599 Chr.17: 41878817-41878817 on Build GRCh38
rs782788159 Chr.17: 41878828-41878828 on Build GRCh38
rs782808207 Chr.17: 41878712-41878712 on Build GRCh38
rs782680537 Chr.17: 41878842-41878843 on Build GRCh38
rs782465452 Chr.17: 41878750-41878750 on Build GRCh38
rs990133806 Chr.17: 41878597-41878597 on Build GRCh38
rs782678424 Chr.17: 41878928-41878928 on Build GRCh38
rs946009376 Chr.17: 41878603-41878603 on Build GRCh38
rs1011460862 Chr.17: 41878887-41878887 on Build GRCh38
rs912601188 Chr.17: 41878657-41878657 on Build GRCh38
rs368177442 Chr.17: 41878769-41878769 on Build GRCh38
rs782119490 Chr.17: 41878827-41878827 on Build GRCh38
rs200877438 Chr.17: 41878851-41878851 on Build GRCh38
rs782277396 Chr.17: 41878802-41878802 on Build GRCh38
rs782331804 Chr.17: 41878944-41878944 on Build GRCh38
rs550130995 Chr.17: 41878696-41878696 on Build GRCh38
rs781784355 Chr.17: 41878764-41878764 on Build GRCh38
rs80121375 Chr.17: 41878606-41878606 on Build GRCh38
rs782744808 Chr.17: 41878808-41878808 on Build GRCh38
rs782367205 Chr.17: 41878773-41878773 on Build GRCh38
rs782620563 Chr.17: 41878760-41878760 on Build GRCh38
rs782537124 Chr.17: 41878896-41878896 on Build GRCh38
rs368407988 Chr.17: 41878878-41878878 on Build GRCh38
rs371847332 Chr.17: 41878893-41878893 on Build GRCh38
rs782066671 Chr.17: 41878805-41878805 on Build GRCh38
rs920213442 Chr.17: 41878697-41878697 on Build GRCh38
rs782585509 Chr.17: 41878770-41878770 on Build GRCh38
rs149943076 Chr.17: 41878961-41878961 on Build GRCh38
rs113569980 Chr.17: 41878591-41878591 on Build GRCh38
rs782144660 Chr.17: 41878700-41878700 on Build GRCh38
rs1048785708 Chr.17: 41878748-41878748 on Build GRCh38
rs781815775 Chr.17: 41878873-41878873 on Build GRCh38
rs782003465 Chr.17: 41878790-41878790 on Build GRCh38
rs367579663 Chr.17: 41878681-41878681 on Build GRCh38
rs371643897 Chr.17: 41878772-41878772 on Build GRCh38
rs942891970 Chr.17: 41878659-41878659 on Build GRCh38
rs886119069 Chr.17: 41878754-41878754 on Build GRCh38
rs781896788 Chr.17: 41878847-41878847 on Build GRCh38
rs781923528 Chr.17: 41878803-41878803 on Build GRCh38

View Full Product Details