The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013703.3 | NP_001013725.2 | 23 - 24 | 3430 | 101 |
XM_005254392.2 | XP_005254449.1 | 23 - 24 | 3453 | 101 | |
XM_011521599.2 | XP_011519901.1 | 23 - 24 | 3453 | 101 | |
XM_011521600.2 | XP_011519902.1 | 23 - 24 | 3453 | 101 | |
XM_017022219.1 | XP_016877708.1 | 23 - 24 | 3453 | 101 | |
GenBank mRNA | AB037759.1 | - | 19 - 20 | 2896 | 101 |
AJ243428.1 | - | 1 - 2 | 57 | 101 | |
AK027011.1 | - | 9 - 10 | 868 | 101 | |
AK074361.1 | - | 2 - 3 | 269 | 101 | |
AL157497.1 | - | 12 - 13 | 1227 | 101 | |
AL832907.1 | - | 17 - 18 | 2690 | 101 | |
BC009350.2 | - | 9 - 10 | 900 | 101 | |
BX649048.1 | - | 6 - 7 | 2275 | 101 |