Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42850004-42850503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOC2
Entrez Gene ID
314
Gene Name
amine oxidase, copper containing 2
Gene Aliases
DAO2, RAO
UniGene
Hs.143102
Gene Chromosome Location
Chr.17: 42844592-42850707 on Build GRCh38
Transcripts
NM_001158.3 NM_009590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346552 c.2171G>T p.G724V Chr.17: 42850248-42850248 on Build GRCh38
1750059 c.2053G>C p.E685Q Chr.17: 42850130-42850130 on Build GRCh38
159104 c.2006A>T p.D669V Chr.17: 42850083-42850083 on Build GRCh38
5942698 c.2063C>T p.P688L Chr.17: 42850140-42850140 on Build GRCh38
3819675 c.2201C>A p.P734H Chr.17: 42850278-42850278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754034436 Chr.17: 42850063-42850065 on Build GRCh38
rs35315871 Chr.17: 42850392-42850393 on Build GRCh38
rs750207298 Chr.17: 42850152-42850153 on Build GRCh38
rs549787218 Chr.17: 42850181-42850181 on Build GRCh38
rs756474151 Chr.17: 42850251-42850251 on Build GRCh38
rs370107506 Chr.17: 42850044-42850044 on Build GRCh38
rs779518566 Chr.17: 42850322-42850322 on Build GRCh38
rs775615898 Chr.17: 42850212-42850212 on Build GRCh38
rs762012675 Chr.17: 42850315-42850315 on Build GRCh38
rs370575506 Chr.17: 42850129-42850129 on Build GRCh38
rs199508507 Chr.17: 42850057-42850057 on Build GRCh38
rs557656858 Chr.17: 42850367-42850367 on Build GRCh38
rs202093818 Chr.17: 42850034-42850034 on Build GRCh38
rs758057836 Chr.17: 42850372-42850372 on Build GRCh38
rs779978308 Chr.17: 42850391-42850391 on Build GRCh38
rs960054528 Chr.17: 42850472-42850472 on Build GRCh38
rs756451050 Chr.17: 42850175-42850175 on Build GRCh38
rs771131465 Chr.17: 42850147-42850148 on Build GRCh38
rs114818002 Chr.17: 42850295-42850295 on Build GRCh38
rs183891874 Chr.17: 42850223-42850223 on Build GRCh38
rs757353278 Chr.17: 42850228-42850228 on Build GRCh38
rs146472486 Chr.17: 42850171-42850171 on Build GRCh38
rs772339214 Chr.17: 42850400-42850400 on Build GRCh38
rs536943629 Chr.17: 42850346-42850346 on Build GRCh38
rs762390295 Chr.17: 42850101-42850101 on Build GRCh38
rs867635527 Chr.17: 42850458-42850458 on Build GRCh38
rs62001928 Chr.17: 42850147-42850147 on Build GRCh38
rs200623048 Chr.17: 42850348-42850348 on Build GRCh38
rs111473367 Chr.17: 42850168-42850168 on Build GRCh38
rs748949528 Chr.17: 42850254-42850254 on Build GRCh38
rs775150527 Chr.17: 42850095-42850095 on Build GRCh38
rs752308610 Chr.17: 42850328-42850328 on Build GRCh38
rs145734216 Chr.17: 42850123-42850123 on Build GRCh38
rs747673953 Chr.17: 42850269-42850269 on Build GRCh38
rs774322251 Chr.17: 42850207-42850207 on Build GRCh38
rs748445445 Chr.17: 42850393-42850393 on Build GRCh38
rs961232942 Chr.17: 42850437-42850437 on Build GRCh38
rs745766844 Chr.17: 42850389-42850389 on Build GRCh38
rs752919084 Chr.17: 42850133-42850133 on Build GRCh38
rs543251149 Chr.17: 42850417-42850417 on Build GRCh38
rs376489968 Chr.17: 42850075-42850075 on Build GRCh38
rs768337675 Chr.17: 42850297-42850297 on Build GRCh38
rs999530053 Chr.17: 42850222-42850222 on Build GRCh38
rs199717908 Chr.17: 42850201-42850201 on Build GRCh38
rs199908351 Chr.17: 42850071-42850071 on Build GRCh38
rs143413007 Chr.17: 42850262-42850262 on Build GRCh38
rs769672296 Chr.17: 42850358-42850358 on Build GRCh38
rs998503360 Chr.17: 42850062-42850062 on Build GRCh38
rs772005140 Chr.17: 42850195-42850195 on Build GRCh38
rs757839016 Chr.17: 42850411-42850415 on Build GRCh38
rs35411053 Chr.17: 42850402-42850402 on Build GRCh38
rs141025432 Chr.17: 42850178-42850178 on Build GRCh38
rs6503720 Chr.17: 42850420-42850420 on Build GRCh38
rs371391603 Chr.17: 42850371-42850371 on Build GRCh38
rs370516177 Chr.17: 42850037-42850037 on Build GRCh38
rs148420019 Chr.17: 42850130-42850130 on Build GRCh38
rs201623521 Chr.17: 42850227-42850227 on Build GRCh38
rs559625786 Chr.17: 42850431-42850431 on Build GRCh38
rs754104348 Chr.17: 42850049-42850049 on Build GRCh38
rs997944272 Chr.17: 42850234-42850234 on Build GRCh38
rs758448935 Chr.17: 42850065-42850065 on Build GRCh38
rs200452450 Chr.17: 42850332-42850332 on Build GRCh38
rs536168385 Chr.17: 42850322-42850322 on Build GRCh38
rs745984959 Chr.17: 42850256-42850256 on Build GRCh38
rs572527050 Chr.17: 42850410-42850410 on Build GRCh38
rs768112479 Chr.17: 42850109-42850109 on Build GRCh38
rs542939436 Chr.17: 42850199-42850199 on Build GRCh38
rs763247499 Chr.17: 42850221-42850221 on Build GRCh38
rs369025380 Chr.17: 42850079-42850079 on Build GRCh38
rs367802604 Chr.17: 42850363-42850363 on Build GRCh38
rs150190092 Chr.17: 42850334-42850334 on Build GRCh38
rs756672081 Chr.17: 42850386-42850386 on Build GRCh38
rs773834689 Chr.17: 42850188-42850188 on Build GRCh38
rs774310558 Chr.17: 42850112-42850112 on Build GRCh38
rs781736600 Chr.17: 42850166-42850166 on Build GRCh38
rs761228616 Chr.17: 42850321-42850321 on Build GRCh38
rs376029073 Chr.17: 42850325-42850325 on Build GRCh38
rs763334115 Chr.17: 42850296-42850296 on Build GRCh38
rs375714451 Chr.17: 42850383-42850383 on Build GRCh38
rs777215660 Chr.17: 42850381-42850381 on Build GRCh38
rs368198240 Chr.17: 42850204-42850204 on Build GRCh38
rs953848524 Chr.17: 42850284-42850284 on Build GRCh38
rs368810599 Chr.17: 42850339-42850339 on Build GRCh38
rs889516277 Chr.17: 42850408-42850408 on Build GRCh38
rs746979351 Chr.17: 42850340-42850340 on Build GRCh38
rs761807846 Chr.17: 42850069-42850071 on Build GRCh38
rs768159974 Chr.17: 42850236-42850236 on Build GRCh38
rs576852504 Chr.17: 42850473-42850473 on Build GRCh38
rs755138332 Chr.17: 42850050-42850050 on Build GRCh38
rs754202494 Chr.17: 42850323-42850323 on Build GRCh38
rs767601829 Chr.17: 42850318-42850318 on Build GRCh38
rs773056749 Chr.17: 42850294-42850294 on Build GRCh38
rs771526592 Chr.17: 42850289-42850289 on Build GRCh38
rs976605093 Chr.17: 42850244-42850244 on Build GRCh38
rs191448188 Chr.17: 42850368-42850368 on Build GRCh38
rs34625494 Chr.17: 42850152-42850152 on Build GRCh38
rs769759130 Chr.17: 42850390-42850390 on Build GRCh38
rs183574353 Chr.17: 42850387-42850387 on Build GRCh38
rs779602703 Chr.17: 42850177-42850177 on Build GRCh38
rs200537033 Chr.17: 42850272-42850272 on Build GRCh38
rs748147749 Chr.17: 42850193-42850193 on Build GRCh38
rs750901007 Chr.17: 42850249-42850249 on Build GRCh38
rs140452679 Chr.17: 42850314-42850314 on Build GRCh38
rs113330937 Chr.17: 42850045-42850045 on Build GRCh38
rs780441544 Chr.17: 42850253-42850253 on Build GRCh38
rs758221237 Chr.17: 42850167-42850167 on Build GRCh38
rs555189786 Chr.17: 42850451-42850451 on Build GRCh38
rs770898058 Chr.17: 42850341-42850341 on Build GRCh38
rs757827651 Chr.17: 42850153-42850153 on Build GRCh38
rs1022937020 Chr.17: 42850479-42850479 on Build GRCh38
rs766919160 Chr.17: 42850370-42850370 on Build GRCh38
rs761714290 Chr.17: 42850117-42850117 on Build GRCh38
rs779119800 Chr.17: 42850053-42850053 on Build GRCh38
rs543218356 Chr.17: 42850138-42850138 on Build GRCh38
rs556549786 Chr.17: 42850364-42850364 on Build GRCh38
rs34587880 Chr.17: 42850321-42850322 on Build GRCh38
rs149660468 Chr.17: 42850093-42850093 on Build GRCh38
rs747480689 Chr.17: 42850073-42850073 on Build GRCh38
rs994095665 Chr.17: 42850450-42850450 on Build GRCh38
rs767355304 Chr.17: 42850118-42850118 on Build GRCh38
rs745592498 Chr.17: 42850083-42850083 on Build GRCh38
rs565401622 Chr.17: 42850267-42850267 on Build GRCh38

View Full Product Details