Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RC3H2
Entrez Gene ID
54542
Gene Name
ring finger and CCCH-type domains 2
Gene Aliases
MNAB, RNF164
Chromosome Location
Chr.9: 122849453 - 122905283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100588.1 NP_001094058.1 7 - 8 1337 71
NM_018835.2 NP_061323.2 7 - 8 1337 71
GenBank mRNA AF255303.1 - 6 - 7 1697 71
AK000308.1 - 5 - 6 771 71
AK226010.1 - 5 - 6 771 71
AL833177.1 - 7 - 8 1276 71
AL834431.1 - 1 - 2 88 71
BC044642.1 - 7 - 8 1269 71

View Full Product Details