Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC16
Entrez Gene ID
23341
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Gene Aliases
-
Chromosome Location
Chr.1: 15526694 - 15571733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015291.3 NP_056106.1 2 - 3 336 96
NR_109898.1 2 - 3 336 96
GenBank mRNA AB023179.2 - 2 - 3 292 96
AK091848.1 - 2 - 3 294 96
AK292479.1 - 2 - 3 268 96
AL833299.1 - 2 - 3 275 96
BC047363.1 - 2 - 3 336 96
BX648982.1 - 2 - 3 261 96

View Full Product Details