The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136051.2 | NP_001129523.1 | 12 - 13 | 1364 | 101 |
NM_153377.4 | NP_700356.2 | 12 - 13 | 1783 | 101 | |
XM_011537893.2 | XP_011536195.1 | 12 - 13 | 1455 | 101 | |
XM_017018790.1 | XP_016874279.1 | 12 - 13 | 1785 | 101 | |
GenBank mRNA | AK074921.1 | - | 3 - 4 | 283 | 101 |
AK292857.1 | - | 12 - 13 | 1764 | 101 | |
AK301162.1 | - | 12 - 13 | 1344 | 101 | |
AL833710.1 | - | 1 - 2 | 120 | 101 | |
AY358288.1 | - | 12 - 13 | 1726 | 101 | |
AY358295.1 | - | 12 - 13 | 1364 | 101 | |
AY505340.1 | - | 12 - 13 | 1534 | 101 | |
BC126169.1 | - | 12 - 13 | 1679 | 101 | |
BC126171.1 | - | 12 - 13 | 1679 | 101 | |
BC143960.1 | - | 12 - 13 | 1679 | 101 |