Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 147043798-147044322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
UniGene
Hs.655684
Gene Chromosome Location
Chr.7: 146116035-148420998 on Build GRCh38
Transcripts
NM_014141.5 XM_017011950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4498089 c.511C>T p.R171C Chr.7: 147044015-147044015 on Build GRCh38
5928909 c.498G>A p.W166* Chr.7: 147044002-147044002 on Build GRCh38
3259315 c.529G>A p.E177K Chr.7: 147044033-147044033 on Build GRCh38
3259312 c.445G>A p.E149K Chr.7: 147043949-147043949 on Build GRCh38
3733058 c.515T>G p.I172S Chr.7: 147044019-147044019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057414305 Chr.7: 147044297-147044297 on Build GRCh38
rs776175685 Chr.7: 147044064-147044064 on Build GRCh38
rs1035729625 Chr.7: 147044175-147044175 on Build GRCh38
rs1042829496 Chr.7: 147044220-147044220 on Build GRCh38
rs1053307930 Chr.7: 147044125-147044125 on Build GRCh38
rs776537172 Chr.7: 147043979-147043979 on Build GRCh38
rs992040536 Chr.7: 147044181-147044181 on Build GRCh38
rs759573683 Chr.7: 147043896-147043896 on Build GRCh38
rs947085301 Chr.7: 147044211-147044211 on Build GRCh38
rs201326295 Chr.7: 147044019-147044019 on Build GRCh38
rs1052026378 Chr.7: 147043823-147043823 on Build GRCh38
rs566125404 Chr.7: 147043864-147043864 on Build GRCh38
rs781338949 Chr.7: 147043940-147043940 on Build GRCh38
rs777481825 Chr.7: 147044042-147044042 on Build GRCh38
rs926546148 Chr.7: 147043844-147043844 on Build GRCh38
rs564259948 Chr.7: 147044262-147044262 on Build GRCh38
rs187423028 Chr.7: 147044171-147044171 on Build GRCh38
rs753713155 Chr.7: 147043890-147043890 on Build GRCh38
rs772931700 Chr.7: 147043974-147043974 on Build GRCh38
rs757977085 Chr.7: 147044026-147044026 on Build GRCh38
rs534910637 Chr.7: 147043934-147043934 on Build GRCh38
rs957156835 Chr.7: 147044183-147044183 on Build GRCh38
rs891925799 Chr.7: 147044127-147044127 on Build GRCh38
rs112651374 Chr.7: 147044177-147044177 on Build GRCh38
rs760487255 Chr.7: 147043874-147043874 on Build GRCh38
rs767592618 Chr.7: 147043998-147043998 on Build GRCh38
rs762423320 Chr.7: 147044203-147044204 on Build GRCh38
rs377060643 Chr.7: 147044161-147044161 on Build GRCh38
rs200736560 Chr.7: 147044090-147044090 on Build GRCh38
rs755330362 Chr.7: 147044018-147044018 on Build GRCh38
rs1053267036 Chr.7: 147043857-147043857 on Build GRCh38
rs575629484 Chr.7: 147044077-147044077 on Build GRCh38
rs759357451 Chr.7: 147043981-147043981 on Build GRCh38
rs746724558 Chr.7: 147043975-147043975 on Build GRCh38
rs937201669 Chr.7: 147044263-147044263 on Build GRCh38
rs202022704 Chr.7: 147044202-147044203 on Build GRCh38
rs780564784 Chr.7: 147044231-147044231 on Build GRCh38
rs565748852 Chr.7: 147043894-147043894 on Build GRCh38
rs1032798348 Chr.7: 147044097-147044097 on Build GRCh38
rs750643318 Chr.7: 147044008-147044008 on Build GRCh38
rs761820697 Chr.7: 147043993-147043993 on Build GRCh38
rs1012175218 Chr.7: 147044145-147044145 on Build GRCh38
rs587780320 Chr.7: 147043989-147043989 on Build GRCh38
rs148378854 Chr.7: 147044201-147044202 on Build GRCh38
rs771891935 Chr.7: 147043866-147043866 on Build GRCh38
rs868783000 Chr.7: 147043913-147043913 on Build GRCh38
rs745625118 Chr.7: 147044069-147044069 on Build GRCh38
rs536003257 Chr.7: 147043838-147043838 on Build GRCh38
rs756407470 Chr.7: 147043951-147043951 on Build GRCh38
rs778256879 Chr.7: 147043968-147043968 on Build GRCh38
rs950502597 Chr.7: 147044280-147044280 on Build GRCh38
rs199851989 Chr.7: 147043953-147043953 on Build GRCh38
rs1004911642 Chr.7: 147044033-147044033 on Build GRCh38
rs796536399 Chr.7: 147044202-147044203 on Build GRCh38
rs368487049 Chr.7: 147044016-147044016 on Build GRCh38
rs748668296 Chr.7: 147044022-147044022 on Build GRCh38
rs769256063 Chr.7: 147043962-147043962 on Build GRCh38
rs939290738 Chr.7: 147043851-147043851 on Build GRCh38
rs765586655 Chr.7: 147044104-147044104 on Build GRCh38
rs775342903 Chr.7: 147044075-147044075 on Build GRCh38
rs747621697 Chr.7: 147043972-147043972 on Build GRCh38
rs866657669 Chr.7: 147043933-147043933 on Build GRCh38
rs138738227 Chr.7: 147043983-147043983 on Build GRCh38
rs750661428 Chr.7: 147043948-147043948 on Build GRCh38
rs746636648 Chr.7: 147044047-147044047 on Build GRCh38
rs577930923 Chr.7: 147043973-147043973 on Build GRCh38
rs767576057 Chr.7: 147044086-147044086 on Build GRCh38
rs988100614 Chr.7: 147044175-147044176 on Build GRCh38
rs1057518820 Chr.7: 147044040-147044040 on Build GRCh38
rs370517200 Chr.7: 147043920-147043920 on Build GRCh38
rs985059232 Chr.7: 147044284-147044284 on Build GRCh38
rs769487232 Chr.7: 147044070-147044070 on Build GRCh38
rs749748111 Chr.7: 147043961-147043961 on Build GRCh38
rs991986215 Chr.7: 147043858-147043858 on Build GRCh38
rs549200396 Chr.7: 147043847-147043847 on Build GRCh38
rs757354647 Chr.7: 147043936-147043936 on Build GRCh38
rs766583902 Chr.7: 147044091-147044091 on Build GRCh38
rs912977248 Chr.7: 147044202-147044205 on Build GRCh38
rs1052828842 Chr.7: 147044198-147044201 on Build GRCh38
rs773332302 Chr.7: 147044087-147044087 on Build GRCh38
rs997756317 Chr.7: 147044119-147044119 on Build GRCh38
rs770653132 Chr.7: 147044050-147044050 on Build GRCh38
rs943684060 Chr.7: 147044192-147044192 on Build GRCh38
rs773031497 Chr.7: 147043871-147043871 on Build GRCh38
rs754031591 Chr.7: 147044093-147044093 on Build GRCh38
rs180726032 Chr.7: 147044102-147044102 on Build GRCh38
rs766280931 Chr.7: 147043876-147043876 on Build GRCh38
rs1021680995 Chr.7: 147044155-147044155 on Build GRCh38
rs770617588 Chr.7: 147043977-147043977 on Build GRCh38
rs796052389 Chr.7: 147044000-147044000 on Build GRCh38
rs747303339 Chr.7: 147044233-147044233 on Build GRCh38
rs111699393 Chr.7: 147044268-147044268 on Build GRCh38
rs375032955 Chr.7: 147044015-147044015 on Build GRCh38

View Full Product Details