Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 147132165-147132667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
UniGene
Hs.655684
Gene Chromosome Location
Chr.7: 146116035-148420998 on Build GRCh38
Transcripts
NM_014141.5 XM_017011950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190190 c.1348+4A>T p.? Chr.7: 147132513-147132513 on Build GRCh38
3635661 c.1094G>A p.S365N Chr.7: 147132255-147132255 on Build GRCh38
6631897 c.1299T>G p.V433V Chr.7: 147132460-147132460 on Build GRCh38
4155647 c.1150C>G p.L384V Chr.7: 147132311-147132311 on Build GRCh38
5492326 c.1298T>C p.V433A Chr.7: 147132459-147132459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200673864 Chr.7: 147132420-147132420 on Build GRCh38
rs577325328 Chr.7: 147132583-147132583 on Build GRCh38
rs368057493 Chr.7: 147132266-147132266 on Build GRCh38
rs746759569 Chr.7: 147132216-147132216 on Build GRCh38
rs1008383932 Chr.7: 147132224-147132224 on Build GRCh38
rs79039458 Chr.7: 147132469-147132469 on Build GRCh38
rs994508015 Chr.7: 147132481-147132481 on Build GRCh38
rs761910664 Chr.7: 147132257-147132257 on Build GRCh38
rs776123229 Chr.7: 147132441-147132441 on Build GRCh38
rs777413625 Chr.7: 147132286-147132286 on Build GRCh38
rs760167340 Chr.7: 147132545-147132546 on Build GRCh38
rs796052373 Chr.7: 147132434-147132434 on Build GRCh38
rs893991199 Chr.7: 147132375-147132375 on Build GRCh38
rs773478974 Chr.7: 147132343-147132343 on Build GRCh38
rs763406385 Chr.7: 147132532-147132532 on Build GRCh38
rs143877693 Chr.7: 147132381-147132381 on Build GRCh38
rs778952446 Chr.7: 147132635-147132635 on Build GRCh38
rs184029281 Chr.7: 147132636-147132636 on Build GRCh38
rs746147361 Chr.7: 147132523-147132523 on Build GRCh38
rs779901891 Chr.7: 147132409-147132409 on Build GRCh38
rs34456867 Chr.7: 147132408-147132408 on Build GRCh38
rs771892342 Chr.7: 147132201-147132201 on Build GRCh38
rs952522529 Chr.7: 147132311-147132311 on Build GRCh38
rs761163668 Chr.7: 147132557-147132557 on Build GRCh38
rs187441681 Chr.7: 147132437-147132437 on Build GRCh38
rs779881849 Chr.7: 147132490-147132490 on Build GRCh38
rs796052372 Chr.7: 147132425-147132425 on Build GRCh38
rs776626910 Chr.7: 147132220-147132220 on Build GRCh38
rs972642774 Chr.7: 147132304-147132304 on Build GRCh38
rs753257558 Chr.7: 147132380-147132380 on Build GRCh38
rs756898951 Chr.7: 147132388-147132388 on Build GRCh38
rs750991896 Chr.7: 147132456-147132456 on Build GRCh38
rs910903599 Chr.7: 147132203-147132203 on Build GRCh38
rs772179690 Chr.7: 147132413-147132413 on Build GRCh38
rs112401840 Chr.7: 147132574-147132574 on Build GRCh38
rs563121849 Chr.7: 147132637-147132637 on Build GRCh38
rs375844898 Chr.7: 147132514-147132514 on Build GRCh38
rs764133173 Chr.7: 147132245-147132245 on Build GRCh38
rs868241984 Chr.7: 147132397-147132397 on Build GRCh38
rs770636056 Chr.7: 147132217-147132217 on Build GRCh38
rs754837081 Chr.7: 147132612-147132612 on Build GRCh38
rs373565604 Chr.7: 147132448-147132448 on Build GRCh38
rs774934085 Chr.7: 147132550-147132550 on Build GRCh38
rs759523461 Chr.7: 147132222-147132222 on Build GRCh38
rs1040699485 Chr.7: 147132240-147132240 on Build GRCh38
rs777201584 Chr.7: 147132382-147132382 on Build GRCh38
rs141439475 Chr.7: 147132301-147132301 on Build GRCh38
rs747014037 Chr.7: 147132504-147132504 on Build GRCh38
rs764457770 Chr.7: 147132547-147132547 on Build GRCh38
rs760868722 Chr.7: 147132349-147132349 on Build GRCh38
rs200282837 Chr.7: 147132522-147132522 on Build GRCh38
rs144970713 Chr.7: 147132501-147132501 on Build GRCh38
rs897785918 Chr.7: 147132473-147132473 on Build GRCh38
rs548409884 Chr.7: 147132327-147132327 on Build GRCh38
rs759910711 Chr.7: 147132436-147132436 on Build GRCh38
rs942340157 Chr.7: 147132230-147132230 on Build GRCh38
rs777017978 Chr.7: 147132431-147132431 on Build GRCh38
rs753860243 Chr.7: 147132498-147132498 on Build GRCh38
rs369718091 Chr.7: 147132424-147132424 on Build GRCh38
rs773218141 Chr.7: 147132418-147132418 on Build GRCh38
rs749205725 Chr.7: 147132496-147132496 on Build GRCh38
rs770257017 Chr.7: 147132527-147132527 on Build GRCh38
rs772760608 Chr.7: 147132205-147132205 on Build GRCh38
rs774451711 Chr.7: 147132249-147132249 on Build GRCh38
rs765778701 Chr.7: 147132377-147132377 on Build GRCh38
rs1003978346 Chr.7: 147132297-147132297 on Build GRCh38
rs56356283 Chr.7: 147132472-147132472 on Build GRCh38
rs371839994 Chr.7: 147132306-147132306 on Build GRCh38
rs910998451 Chr.7: 147132314-147132314 on Build GRCh38
rs73471053 Chr.7: 147132280-147132280 on Build GRCh38
rs796052370 Chr.7: 147132334-147132334 on Build GRCh38
rs796052371 Chr.7: 147132399-147132399 on Build GRCh38
rs191933423 Chr.7: 147132544-147132544 on Build GRCh38
rs756812096 Chr.7: 147132457-147132457 on Build GRCh38
rs760012888 Chr.7: 147132361-147132361 on Build GRCh38
rs976492311 Chr.7: 147132597-147132597 on Build GRCh38
rs78543192 Chr.7: 147132298-147132298 on Build GRCh38
rs751202112 Chr.7: 147132383-147132383 on Build GRCh38
rs1052681268 Chr.7: 147132374-147132374 on Build GRCh38
rs745611779 Chr.7: 147132406-147132406 on Build GRCh38
rs375172684 Chr.7: 147132326-147132326 on Build GRCh38
rs763419157 Chr.7: 147132443-147132443 on Build GRCh38
rs771022281 Chr.7: 147132426-147132426 on Build GRCh38
rs751589771 Chr.7: 147132299-147132299 on Build GRCh38
rs750006152 Chr.7: 147132209-147132210 on Build GRCh38
rs922281978 Chr.7: 147132601-147132601 on Build GRCh38
rs755129419 Chr.7: 147132322-147132322 on Build GRCh38
rs755900240 Chr.7: 147132486-147132486 on Build GRCh38
rs935590800 Chr.7: 147132261-147132261 on Build GRCh38
rs796052374 Chr.7: 147132507-147132507 on Build GRCh38
rs537991819 Chr.7: 147132455-147132455 on Build GRCh38
rs775430816 Chr.7: 147132323-147132323 on Build GRCh38
rs147815978 Chr.7: 147132410-147132410 on Build GRCh38
rs766747399 Chr.7: 147132355-147132355 on Build GRCh38
rs149576292 Chr.7: 147132279-147132279 on Build GRCh38
rs776037369 Chr.7: 147132530-147132530 on Build GRCh38
rs755592359 Chr.7: 147132277-147132277 on Build GRCh38
rs1021975064 Chr.7: 147132571-147132571 on Build GRCh38
rs926413848 Chr.7: 147132613-147132613 on Build GRCh38
rs750626971 Chr.7: 147132262-147132262 on Build GRCh38
rs983846593 Chr.7: 147132313-147132313 on Build GRCh38
rs754297905 Chr.7: 147132358-147132358 on Build GRCh38
rs754356364 Chr.7: 147132270-147132270 on Build GRCh38
rs564865065 Chr.7: 147132260-147132260 on Build GRCh38
rs901463412 Chr.7: 147132256-147132256 on Build GRCh38
rs983941500 Chr.7: 147132548-147132548 on Build GRCh38
rs757993314 Chr.7: 147132284-147132284 on Build GRCh38
rs956320440 Chr.7: 147132497-147132497 on Build GRCh38

View Full Product Details