Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151035803-151036300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB8
Entrez Gene ID
11194
Gene Name
ATP binding cassette subfamily B member 8
Gene Aliases
EST328128, M-ABC1, MABC1
UniGene
Hs.647118
Gene Chromosome Location
Chr.7: 151028422-151047782 on Build GRCh38
Transcripts
NM_001282291.1 NM_001282292.1 NM_001282293.1 NM_007188.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3879337 c.736C>T p.Q246* Chr.7: 151035954-151035954 on Build GRCh38
3879336 c.1000C>T p.Q334* Chr.7: 151035954-151035954 on Build GRCh38
4360848 c.1099T>G p.F367V Chr.7: 151035942-151035942 on Build GRCh38
6438403 c.1187_1188CC>TT p.A396V Chr.7: 151036135-151036136 on Build GRCh38
6177210 c.693G>T p.E231D Chr.7: 151035911-151035911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750804178 Chr.7: 151035909-151035909 on Build GRCh38
rs563099517 Chr.7: 151036103-151036103 on Build GRCh38
rs770304156 Chr.7: 151035957-151035957 on Build GRCh38
rs756142033 Chr.7: 151036190-151036190 on Build GRCh38
rs757943876 Chr.7: 151036009-151036009 on Build GRCh38
rs1022171182 Chr.7: 151035896-151035896 on Build GRCh38
rs777741819 Chr.7: 151035945-151035945 on Build GRCh38
rs752310569 Chr.7: 151036105-151036105 on Build GRCh38
rs372274457 Chr.7: 151035838-151035838 on Build GRCh38
rs373829198 Chr.7: 151035898-151035898 on Build GRCh38
rs781358894 Chr.7: 151036216-151036216 on Build GRCh38
rs371323931 Chr.7: 151036073-151036073 on Build GRCh38
rs750205998 Chr.7: 151036122-151036122 on Build GRCh38
rs760124784 Chr.7: 151036142-151036142 on Build GRCh38
rs757211280 Chr.7: 151036111-151036111 on Build GRCh38
rs748204050 Chr.7: 151036075-151036075 on Build GRCh38
rs781046116 Chr.7: 151036121-151036121 on Build GRCh38
rs1029731177 Chr.7: 151035889-151035889 on Build GRCh38
rs748682978 Chr.7: 151035909-151035911 on Build GRCh38
rs76225344 Chr.7: 151036028-151036028 on Build GRCh38
rs900481365 Chr.7: 151035839-151035839 on Build GRCh38
rs1038478089 Chr.7: 151036102-151036102 on Build GRCh38
rs2303926 Chr.7: 151035938-151035938 on Build GRCh38
rs771410792 Chr.7: 151036095-151036095 on Build GRCh38
rs746332866 Chr.7: 151036256-151036257 on Build GRCh38
rs779452010 Chr.7: 151035846-151035866 on Build GRCh38
rs143943108 Chr.7: 151036161-151036161 on Build GRCh38
rs768110885 Chr.7: 151035967-151035967 on Build GRCh38
rs752185748 Chr.7: 151036008-151036008 on Build GRCh38
rs371589693 Chr.7: 151035984-151035984 on Build GRCh38
rs751033196 Chr.7: 151036109-151036109 on Build GRCh38
rs747081126 Chr.7: 151036208-151036208 on Build GRCh38
rs922500393 Chr.7: 151036006-151036006 on Build GRCh38
rs544222964 Chr.7: 151035825-151035845 on Build GRCh38
rs76802349 Chr.7: 151036184-151036184 on Build GRCh38
rs748330890 Chr.7: 151036127-151036127 on Build GRCh38
rs6968942 Chr.7: 151035845-151035845 on Build GRCh38
rs777044789 Chr.7: 151036101-151036101 on Build GRCh38
rs780971233 Chr.7: 151036036-151036036 on Build GRCh38
rs770720559 Chr.7: 151035948-151035948 on Build GRCh38
rs200906042 Chr.7: 151035966-151035966 on Build GRCh38
rs267601419 Chr.7: 151036156-151036156 on Build GRCh38
rs980423131 Chr.7: 151036004-151036006 on Build GRCh38
rs371242848 Chr.7: 151035868-151035868 on Build GRCh38
rs761114613 Chr.7: 151035981-151035981 on Build GRCh38
rs746901227 Chr.7: 151036027-151036027 on Build GRCh38
rs773601458 Chr.7: 151036014-151036026 on Build GRCh38
rs754658023 Chr.7: 151036001-151036001 on Build GRCh38
rs777908393 Chr.7: 151036207-151036207 on Build GRCh38
rs1053555852 Chr.7: 151036169-151036169 on Build GRCh38
rs755489368 Chr.7: 151035842-151035842 on Build GRCh38
rs569927391 Chr.7: 151035985-151035985 on Build GRCh38
rs778422625 Chr.7: 151035937-151035937 on Build GRCh38
rs187662799 Chr.7: 151035884-151035884 on Build GRCh38
rs774567306 Chr.7: 151035960-151035960 on Build GRCh38
rs370026207 Chr.7: 151036162-151036162 on Build GRCh38
rs770877051 Chr.7: 151036209-151036209 on Build GRCh38
rs769885003 Chr.7: 151035882-151035882 on Build GRCh38
rs28496243 Chr.7: 151035866-151035866 on Build GRCh38
rs891887561 Chr.7: 151036048-151036048 on Build GRCh38
rs543912073 Chr.7: 151036052-151036052 on Build GRCh38
rs377214975 Chr.7: 151036131-151036131 on Build GRCh38
rs141000427 Chr.7: 151036012-151036012 on Build GRCh38
rs761183737 Chr.7: 151036094-151036094 on Build GRCh38
rs1140980 Chr.7: 151035944-151035944 on Build GRCh38
rs369491387 Chr.7: 151036023-151036023 on Build GRCh38
rs951148869 Chr.7: 151036182-151036182 on Build GRCh38
rs775849676 Chr.7: 151036160-151036160 on Build GRCh38
rs771684717 Chr.7: 151036178-151036178 on Build GRCh38
rs772235389 Chr.7: 151036078-151036078 on Build GRCh38
rs201786514 Chr.7: 151035908-151035908 on Build GRCh38
rs755902689 Chr.7: 151036125-151036125 on Build GRCh38
rs763118021 Chr.7: 151035885-151035885 on Build GRCh38
rs774585394 Chr.7: 151035899-151035899 on Build GRCh38
rs367985731 Chr.7: 151035935-151035935 on Build GRCh38
rs565619420 Chr.7: 151036128-151036128 on Build GRCh38
rs754606258 Chr.7: 151035936-151035936 on Build GRCh38
rs768763784 Chr.7: 151035959-151035959 on Build GRCh38
rs760525069 Chr.7: 151035876-151035876 on Build GRCh38
rs200124806 Chr.7: 151036107-151036107 on Build GRCh38
rs750287884 Chr.7: 151036186-151036186 on Build GRCh38
rs147045076 Chr.7: 151035928-151035928 on Build GRCh38
rs975265890 Chr.7: 151036239-151036239 on Build GRCh38
rs771448001 Chr.7: 151036134-151036134 on Build GRCh38
rs772359324 Chr.7: 151035994-151035996 on Build GRCh38
rs772005013 Chr.7: 151035856-151035856 on Build GRCh38
rs773892521 Chr.7: 151035977-151035977 on Build GRCh38
rs768908560 Chr.7: 151036063-151036063 on Build GRCh38
rs376741483 Chr.7: 151036200-151036200 on Build GRCh38
rs148169229 Chr.7: 151036133-151036133 on Build GRCh38
rs768204790 Chr.7: 151035904-151035904 on Build GRCh38
rs756591786 Chr.7: 151035923-151035923 on Build GRCh38
rs773445092 Chr.7: 151036085-151036085 on Build GRCh38
rs376146863 Chr.7: 151036013-151036013 on Build GRCh38
rs779935327 Chr.7: 151036198-151036198 on Build GRCh38
rs1140979 Chr.7: 151035881-151035881 on Build GRCh38
rs749467318 Chr.7: 151035958-151035958 on Build GRCh38
rs144862157 Chr.7: 151036126-151036126 on Build GRCh38
rs202226613 Chr.7: 151036201-151036201 on Build GRCh38
rs537104835 Chr.7: 151036277-151036277 on Build GRCh38
rs751063378 Chr.7: 151036173-151036173 on Build GRCh38
rs138257786 Chr.7: 151035947-151035947 on Build GRCh38
rs777044246 Chr.7: 151036138-151036138 on Build GRCh38
rs770099809 Chr.7: 151036157-151036157 on Build GRCh38
rs774763369 Chr.7: 151036074-151036074 on Build GRCh38
rs200911168 Chr.7: 151035849-151035849 on Build GRCh38
rs747214277 Chr.7: 151036060-151036061 on Build GRCh38
rs142475735 Chr.7: 151035900-151035900 on Build GRCh38
rs374885136 Chr.7: 151036108-151036108 on Build GRCh38

View Full Product Details