Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30614221-30614767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2E2
Entrez Gene ID
2961
Gene Name
general transcription factor IIE subunit 2
Gene Aliases
FE, TF2E2, TFIIE-B, TTD6
UniGene
Hs.77100
Gene Chromosome Location
Chr.8: 30578514-30658241 on Build GRCh38
Transcripts
NM_002095.4 XM_011544509.2 XM_011544510.1 XM_017013363.1 XM_017013364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
261210 c.366+7A>C p.? Chr.8: 30614601-30614601 on Build GRCh38
454438 c.259-1G>A p.? Chr.8: 30614716-30614716 on Build GRCh38
1241236 c.272G>A p.R91Q Chr.8: 30614702-30614702 on Build GRCh38
5029665 c.282G>A p.T94T Chr.8: 30614692-30614692 on Build GRCh38
5785173 c.319C>T p.Q107* Chr.8: 30614655-30614655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972870606 Chr.8: 30614447-30614447 on Build GRCh38
rs751095510 Chr.8: 30614704-30614704 on Build GRCh38
rs971239026 Chr.8: 30614405-30614405 on Build GRCh38
rs777646389 Chr.8: 30614697-30614697 on Build GRCh38
rs902064993 Chr.8: 30614486-30614486 on Build GRCh38
rs746850928 Chr.8: 30614707-30614707 on Build GRCh38
rs971896773 Chr.8: 30614709-30614709 on Build GRCh38
rs372701593 Chr.8: 30614583-30614583 on Build GRCh38
rs768289418 Chr.8: 30614710-30614710 on Build GRCh38
rs762536890 Chr.8: 30614645-30614645 on Build GRCh38
rs541510806 Chr.8: 30614376-30614376 on Build GRCh38
rs552879603 Chr.8: 30614479-30614479 on Build GRCh38
rs754124140 Chr.8: 30614703-30614703 on Build GRCh38
rs370136395 Chr.8: 30614265-30614265 on Build GRCh38
rs1034137133 Chr.8: 30614365-30614365 on Build GRCh38
rs930343216 Chr.8: 30614418-30614418 on Build GRCh38
rs28735275 Chr.8: 30614371-30614371 on Build GRCh38
rs998606339 Chr.8: 30614345-30614345 on Build GRCh38
rs772924568 Chr.8: 30614630-30614630 on Build GRCh38
rs546794521 Chr.8: 30614443-30614443 on Build GRCh38
rs182600571 Chr.8: 30614587-30614587 on Build GRCh38
rs888812626 Chr.8: 30614304-30614304 on Build GRCh38
rs910202748 Chr.8: 30614480-30614480 on Build GRCh38
rs1042494457 Chr.8: 30614520-30614520 on Build GRCh38
rs769323215 Chr.8: 30614397-30614397 on Build GRCh38
rs751991227 Chr.8: 30614656-30614656 on Build GRCh38
rs114292437 Chr.8: 30614676-30614676 on Build GRCh38
rs1024658169 Chr.8: 30614702-30614702 on Build GRCh38
rs1040447545 Chr.8: 30614517-30614517 on Build GRCh38
rs766007320 Chr.8: 30614651-30614651 on Build GRCh38
rs1021007851 Chr.8: 30614300-30614300 on Build GRCh38
rs575971465 Chr.8: 30614346-30614346 on Build GRCh38
rs757783461 Chr.8: 30614558-30614558 on Build GRCh38
rs919020499 Chr.8: 30614416-30614416 on Build GRCh38
rs34835819 Chr.8: 30614540-30614541 on Build GRCh38
rs771045931 Chr.8: 30614310-30614310 on Build GRCh38
rs779271698 Chr.8: 30614705-30614705 on Build GRCh38
rs961454645 Chr.8: 30614437-30614437 on Build GRCh38
rs768756005 Chr.8: 30614607-30614607 on Build GRCh38
rs770606981 Chr.8: 30614735-30614735 on Build GRCh38
rs774826032 Chr.8: 30614462-30614462 on Build GRCh38
rs555065014 Chr.8: 30614616-30614616 on Build GRCh38
rs1024207978 Chr.8: 30614403-30614403 on Build GRCh38
rs745406623 Chr.8: 30614379-30614379 on Build GRCh38
rs773966002 Chr.8: 30614738-30614738 on Build GRCh38
rs1048735938 Chr.8: 30614282-30614282 on Build GRCh38
rs749480638 Chr.8: 30614595-30614595 on Build GRCh38
rs11781969 Chr.8: 30614295-30614295 on Build GRCh38
rs114889473 Chr.8: 30614317-30614317 on Build GRCh38
rs764697154 Chr.8: 30614585-30614586 on Build GRCh38
rs753094265 Chr.8: 30614692-30614692 on Build GRCh38
rs771996510 Chr.8: 30614596-30614596 on Build GRCh38
rs369034600 Chr.8: 30614720-30614720 on Build GRCh38
rs374079613 Chr.8: 30614391-30614391 on Build GRCh38
rs892994715 Chr.8: 30614708-30614708 on Build GRCh38
rs1031473776 Chr.8: 30614355-30614355 on Build GRCh38
rs367983000 Chr.8: 30614452-30614452 on Build GRCh38
rs530205020 Chr.8: 30614382-30614382 on Build GRCh38
rs532685346 Chr.8: 30614471-30614471 on Build GRCh38
rs762449060 Chr.8: 30614733-30614733 on Build GRCh38
rs755301412 Chr.8: 30614671-30614671 on Build GRCh38
rs985760529 Chr.8: 30614478-30614478 on Build GRCh38
rs533926333 Chr.8: 30614712-30614712 on Build GRCh38
rs1035816289 Chr.8: 30614414-30614414 on Build GRCh38
rs561198137 Chr.8: 30614547-30614548 on Build GRCh38
rs943089793 Chr.8: 30614493-30614493 on Build GRCh38
rs775365025 Chr.8: 30614597-30614597 on Build GRCh38
rs572070074 Chr.8: 30614355-30614355 on Build GRCh38
rs555914396 Chr.8: 30614473-30614473 on Build GRCh38
rs901559501 Chr.8: 30614548-30614548 on Build GRCh38
rs781181967 Chr.8: 30614339-30614339 on Build GRCh38
rs764614723 Chr.8: 30614623-30614623 on Build GRCh38
rs150253810 Chr.8: 30614377-30614377 on Build GRCh38
rs776412409 Chr.8: 30614612-30614612 on Build GRCh38
rs756120809 Chr.8: 30614576-30614576 on Build GRCh38
rs372567210 Chr.8: 30614732-30614732 on Build GRCh38
rs370799297 Chr.8: 30614598-30614598 on Build GRCh38
rs545740705 Chr.8: 30614306-30614306 on Build GRCh38
rs541419495 Chr.8: 30614375-30614376 on Build GRCh38

View Full Product Details