Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31066901-31067429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7254536 c.565C>A p.L189I Chr.8: 31067093-31067093 on Build GRCh38
4397965 c.581C>T p.S194F Chr.8: 31067109-31067109 on Build GRCh38
5367737 c.606_607insT p.P204fs*4 Chr.8: 31067134-31067135 on Build GRCh38
5903691 c.610C>T p.P204S Chr.8: 31067138-31067138 on Build GRCh38
73303 c.631C>G p.L211V Chr.8: 31067159-31067159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016545658 Chr.8: 31066927-31066927 on Build GRCh38
rs773262209 Chr.8: 31067216-31067216 on Build GRCh38
rs927350912 Chr.8: 31067113-31067113 on Build GRCh38
rs758515093 Chr.8: 31067215-31067216 on Build GRCh38
rs554174469 Chr.8: 31067372-31067372 on Build GRCh38
rs930667475 Chr.8: 31067293-31067293 on Build GRCh38
rs770545798 Chr.8: 31067064-31067064 on Build GRCh38
rs375762379 Chr.8: 31067114-31067114 on Build GRCh38
rs776831463 Chr.8: 31067109-31067109 on Build GRCh38
rs1008491153 Chr.8: 31067345-31067345 on Build GRCh38
rs11574205 Chr.8: 31067317-31067317 on Build GRCh38
rs746210769 Chr.8: 31067076-31067076 on Build GRCh38
rs996483683 Chr.8: 31067341-31067341 on Build GRCh38
rs748976035 Chr.8: 31067055-31067055 on Build GRCh38
rs919269017 Chr.8: 31067262-31067262 on Build GRCh38
rs1049081338 Chr.8: 31067299-31067299 on Build GRCh38
rs532897847 Chr.8: 31067264-31067264 on Build GRCh38
rs767516095 Chr.8: 31067129-31067129 on Build GRCh38
rs569128549 Chr.8: 31067344-31067344 on Build GRCh38
rs558866038 Chr.8: 31066955-31066955 on Build GRCh38
rs761918478 Chr.8: 31067195-31067195 on Build GRCh38
rs367991517 Chr.8: 31067042-31067042 on Build GRCh38
rs778337067 Chr.8: 31066978-31066978 on Build GRCh38
rs776917867 Chr.8: 31067187-31067187 on Build GRCh38
rs371736282 Chr.8: 31067111-31067111 on Build GRCh38
rs780927874 Chr.8: 31067025-31067025 on Build GRCh38
rs756050193 Chr.8: 31067141-31067141 on Build GRCh38
rs190475593 Chr.8: 31066940-31066940 on Build GRCh38
rs766327723 Chr.8: 31067004-31067004 on Build GRCh38
rs1007764190 Chr.8: 31066977-31066977 on Build GRCh38
rs980721395 Chr.8: 31067381-31067381 on Build GRCh38
rs972532045 Chr.8: 31067199-31067199 on Build GRCh38
rs1051111254 Chr.8: 31067342-31067342 on Build GRCh38
rs772637078 Chr.8: 31067077-31067077 on Build GRCh38
rs574086493 Chr.8: 31066993-31066993 on Build GRCh38
rs777437332 Chr.8: 31067043-31067043 on Build GRCh38
rs754062727 Chr.8: 31066986-31066986 on Build GRCh38
rs753330899 Chr.8: 31067154-31067154 on Build GRCh38
rs201870156 Chr.8: 31067193-31067193 on Build GRCh38
rs1037904871 Chr.8: 31067288-31067288 on Build GRCh38
rs940853459 Chr.8: 31067250-31067250 on Build GRCh38
rs775802030 Chr.8: 31067089-31067089 on Build GRCh38
rs747520883 Chr.8: 31067182-31067182 on Build GRCh38
rs561603992 Chr.8: 31067115-31067115 on Build GRCh38
rs1040565074 Chr.8: 31067357-31067357 on Build GRCh38
rs956339032 Chr.8: 31067092-31067092 on Build GRCh38
rs759197788 Chr.8: 31067116-31067116 on Build GRCh38
rs777311042 Chr.8: 31067147-31067147 on Build GRCh38
rs760829035 Chr.8: 31067103-31067103 on Build GRCh38
rs1800389 Chr.8: 31067041-31067041 on Build GRCh38
rs374395940 Chr.8: 31067009-31067009 on Build GRCh38
rs138037385 Chr.8: 31067156-31067156 on Build GRCh38
rs938648168 Chr.8: 31067160-31067160 on Build GRCh38
rs76337727 Chr.8: 31067398-31067398 on Build GRCh38
rs568783551 Chr.8: 31067362-31067362 on Build GRCh38
rs538000653 Chr.8: 31067350-31067350 on Build GRCh38
rs778470570 Chr.8: 31067169-31067169 on Build GRCh38
rs907937476 Chr.8: 31067185-31067185 on Build GRCh38
rs577277915 Chr.8: 31067380-31067380 on Build GRCh38
rs114641318 Chr.8: 31067392-31067392 on Build GRCh38
rs771397499 Chr.8: 31067104-31067104 on Build GRCh38
rs199644081 Chr.8: 31067138-31067138 on Build GRCh38
rs570330577 Chr.8: 31067210-31067210 on Build GRCh38
rs992800740 Chr.8: 31067397-31067397 on Build GRCh38
rs554971532 Chr.8: 31067351-31067351 on Build GRCh38
rs574825204 Chr.8: 31067393-31067393 on Build GRCh38
rs765499420 Chr.8: 31067015-31067016 on Build GRCh38
rs746256937 Chr.8: 31067173-31067173 on Build GRCh38
rs764227749 Chr.8: 31067107-31067107 on Build GRCh38
rs777153045 Chr.8: 31067215-31067215 on Build GRCh38
rs576837409 Chr.8: 31067029-31067029 on Build GRCh38
rs756966886 Chr.8: 31067163-31067163 on Build GRCh38
rs772273549 Chr.8: 31067174-31067174 on Build GRCh38
rs780640173 Chr.8: 31067181-31067181 on Build GRCh38
rs980659335 Chr.8: 31067159-31067159 on Build GRCh38
rs534207791 Chr.8: 31067365-31067365 on Build GRCh38
rs752830087 Chr.8: 31067052-31067052 on Build GRCh38
rs769275170 Chr.8: 31067186-31067186 on Build GRCh38
rs774751582 Chr.8: 31067070-31067070 on Build GRCh38

View Full Product Details