Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71096792-71097300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO4
Entrez Gene ID
4303
Gene Name
forkhead box O4
Gene Aliases
AFX, AFX1, MLLT7
UniGene
Hs.584654
Gene Chromosome Location
Chr.X: 71096149-71103534 on Build GRCh38
Transcripts
NM_001170931.1 NM_005938.3

SNP Detail

SNP ID Chromosome Location
rs12013673 Chr.X: 71097249-71097249 on Build GRCh38
rs768812217 Chr.X: 71096985-71096985 on Build GRCh38
rs748394131 Chr.X: 71097001-71097001 on Build GRCh38
rs926871627 Chr.X: 71097164-71097164 on Build GRCh38
rs757757724 Chr.X: 71097089-71097089 on Build GRCh38
rs922417531 Chr.X: 71096815-71096815 on Build GRCh38
rs1056663926 Chr.X: 71097253-71097253 on Build GRCh38
rs771810383 Chr.X: 71096912-71096912 on Build GRCh38
rs749130867 Chr.X: 71096913-71096913 on Build GRCh38
rs753869002 Chr.X: 71097083-71097083 on Build GRCh38
rs755873760 Chr.X: 71096881-71096881 on Build GRCh38
rs371282845 Chr.X: 71096989-71096989 on Build GRCh38
rs979053384 Chr.X: 71097053-71097053 on Build GRCh38
rs977844229 Chr.X: 71096926-71096926 on Build GRCh38
rs772968101 Chr.X: 71097147-71097147 on Build GRCh38
rs938213752 Chr.X: 71097226-71097226 on Build GRCh38
rs756848241 Chr.X: 71096876-71096876 on Build GRCh38
rs750286710 Chr.X: 71096878-71096878 on Build GRCh38
rs1056659902 Chr.X: 71097149-71097149 on Build GRCh38
rs751220339 Chr.X: 71096846-71096846 on Build GRCh38
rs368327326 Chr.X: 71097000-71097000 on Build GRCh38
rs988285482 Chr.X: 71096992-71096992 on Build GRCh38
rs374774363 Chr.X: 71097009-71097009 on Build GRCh38
rs773561538 Chr.X: 71097030-71097030 on Build GRCh38
rs946373171 Chr.X: 71097033-71097033 on Build GRCh38
rs369897569 Chr.X: 71097259-71097259 on Build GRCh38

View Full Product Details