Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71103159-71103654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO4
Entrez Gene ID
4303
Gene Name
forkhead box O4
Gene Aliases
AFX, AFX1, MLLT7
UniGene
Hs.584654
Gene Chromosome Location
Chr.X: 71096149-71103534 on Build GRCh38
Transcripts
NM_001170931.1 NM_005938.3 XM_011530957.2

SNP Detail

SNP ID Chromosome Location
rs1019932719 Chr.X: 71103391-71103391 on Build GRCh38
rs776180620 Chr.X: 71103219-71103219 on Build GRCh38
rs911066460 Chr.X: 71103628-71103628 on Build GRCh38
rs933419786 Chr.X: 71103583-71103583 on Build GRCh38
rs948904911 Chr.X: 71103436-71103436 on Build GRCh38
rs192176526 Chr.X: 71103187-71103187 on Build GRCh38
rs985937738 Chr.X: 71103196-71103196 on Build GRCh38
rs559205115 Chr.X: 71103286-71103286 on Build GRCh38
rs901962583 Chr.X: 71103217-71103217 on Build GRCh38
rs1050929231 Chr.X: 71103247-71103247 on Build GRCh38
rs940948881 Chr.X: 71103549-71103549 on Build GRCh38
rs1019118204 Chr.X: 71103201-71103201 on Build GRCh38
rs958824434 Chr.X: 71103502-71103502 on Build GRCh38
rs923284006 Chr.X: 71103578-71103578 on Build GRCh38
rs916123088 Chr.X: 71103416-71103416 on Build GRCh38
rs972497660 Chr.X: 71103572-71103572 on Build GRCh38
rs184012791 Chr.X: 71103403-71103403 on Build GRCh38
rs985014169 Chr.X: 71103542-71103542 on Build GRCh38
rs1006815376 Chr.X: 71103284-71103284 on Build GRCh38
rs942545244 Chr.X: 71103629-71103629 on Build GRCh38
rs4503258 Chr.X: 71103387-71103387 on Build GRCh38
rs889601076 Chr.X: 71103265-71103265 on Build GRCh38
rs1050461854 Chr.X: 71103593-71103593 on Build GRCh38
rs1036356273 Chr.X: 71103204-71103204 on Build GRCh38
rs977510070 Chr.X: 71103283-71103284 on Build GRCh38
rs893046381 Chr.X: 71103379-71103379 on Build GRCh38
rs909501508 Chr.X: 71103544-71103544 on Build GRCh38
rs34904436 Chr.X: 71103377-71103377 on Build GRCh38
rs1034496136 Chr.X: 71103439-71103439 on Build GRCh38
rs898488151 Chr.X: 71103598-71103598 on Build GRCh38
rs933402074 Chr.X: 71103232-71103232 on Build GRCh38
rs766125604 Chr.X: 71103447-71103447 on Build GRCh38
rs764505779 Chr.X: 71103340-71103342 on Build GRCh38
rs76848997 Chr.X: 71103499-71103499 on Build GRCh38
rs1045861909 Chr.X: 71103441-71103441 on Build GRCh38
rs971585694 Chr.X: 71103402-71103402 on Build GRCh38

View Full Product Details