Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72351591-72352093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC8
Entrez Gene ID
55869
Gene Name
histone deacetylase 8
Gene Aliases
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
UniGene
Hs.310536
Gene Chromosome Location
Chr.X: 72329516-72573103 on Build GRCh38
Transcripts
NM_001166418.1 NM_018486.2 NR_051952.1 XM_011530986.2 XM_017029640.1 XM_017029641.1 XM_017029642.1 XM_017029643.1 XM_017029644.1 XM_017029645.1 XM_017029646.1

SNP Detail

SNP ID Chromosome Location
rs1039038766 Chr.X: 72351717-72351717 on Build GRCh38
rs781979722 Chr.X: 72351760-72351760 on Build GRCh38
rs782244344 Chr.X: 72351887-72351887 on Build GRCh38
rs782463393 Chr.X: 72351812-72351812 on Build GRCh38
rs863224877 Chr.X: 72351840-72351840 on Build GRCh38
rs782357452 Chr.X: 72351694-72351694 on Build GRCh38
rs373892213 Chr.X: 72351723-72351723 on Build GRCh38
rs782617971 Chr.X: 72351831-72351831 on Build GRCh38
rs189038245 Chr.X: 72351908-72351908 on Build GRCh38
rs782581865 Chr.X: 72352066-72352066 on Build GRCh38
rs372334014 Chr.X: 72351727-72351727 on Build GRCh38
rs782717694 Chr.X: 72351777-72351777 on Build GRCh38
rs782752785 Chr.X: 72351755-72351755 on Build GRCh38
rs781894758 Chr.X: 72351683-72351683 on Build GRCh38
rs781988247 Chr.X: 72351712-72351712 on Build GRCh38
rs782418663 Chr.X: 72351703-72351703 on Build GRCh38
rs374293466 Chr.X: 72351761-72351761 on Build GRCh38
rs899676887 Chr.X: 72351995-72351995 on Build GRCh38
rs782180141 Chr.X: 72351841-72351841 on Build GRCh38
rs180782465 Chr.X: 72352067-72352067 on Build GRCh38
rs782687720 Chr.X: 72351873-72351873 on Build GRCh38
rs1011464257 Chr.X: 72352009-72352009 on Build GRCh38
rs781918096 Chr.X: 72351700-72351700 on Build GRCh38
rs112537722 Chr.X: 72352055-72352055 on Build GRCh38
rs781864686 Chr.X: 72351798-72351798 on Build GRCh38
rs782220515 Chr.X: 72351969-72351969 on Build GRCh38
rs368139150 Chr.X: 72351724-72351724 on Build GRCh38
rs782505730 Chr.X: 72351778-72351778 on Build GRCh38
rs782163825 Chr.X: 72351724-72351724 on Build GRCh38
rs782034060 Chr.X: 72351948-72351948 on Build GRCh38
rs370568736 Chr.X: 72351926-72351926 on Build GRCh38
rs147459925 Chr.X: 72351994-72351994 on Build GRCh38
rs782282933 Chr.X: 72351701-72351701 on Build GRCh38
rs781862582 Chr.X: 72351762-72351762 on Build GRCh38
rs782558250 Chr.X: 72351792-72351792 on Build GRCh38
rs782747900 Chr.X: 72351797-72351797 on Build GRCh38
rs782404971 Chr.X: 72351931-72351931 on Build GRCh38
rs370177443 Chr.X: 72351688-72351688 on Build GRCh38
rs782212395 Chr.X: 72351693-72351693 on Build GRCh38
rs782450189 Chr.X: 72351842-72351842 on Build GRCh38
rs1046797566 Chr.X: 72351877-72351877 on Build GRCh38
rs367674943 Chr.X: 72351684-72351684 on Build GRCh38
rs377116815 Chr.X: 72351800-72351800 on Build GRCh38
rs919576396 Chr.X: 72351730-72351730 on Build GRCh38

View Full Product Details