Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72464481-72464982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC8
Entrez Gene ID
55869
Gene Name
histone deacetylase 8
Gene Aliases
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
UniGene
Hs.310536
Gene Chromosome Location
Chr.X: 72329516-72573103 on Build GRCh38
Transcripts
NM_001166418.1 NM_018486.2 NR_051952.1 XM_011530986.2 XM_011530987.2 XM_017029640.1 XM_017029641.1 XM_017029642.1 XM_017029643.1 XM_017029644.1 XM_017029645.1 XM_017029646.1

SNP Detail

SNP ID Chromosome Location
rs782196784 Chr.X: 72464524-72464524 on Build GRCh38
rs782469454 Chr.X: 72464749-72464749 on Build GRCh38
rs782142009 Chr.X: 72464611-72464611 on Build GRCh38
rs976983859 Chr.X: 72464888-72464888 on Build GRCh38
rs782384793 Chr.X: 72464551-72464551 on Build GRCh38
rs782676451 Chr.X: 72464542-72464542 on Build GRCh38
rs782662702 Chr.X: 72464751-72464751 on Build GRCh38
rs961275191 Chr.X: 72464858-72464858 on Build GRCh38
rs953904622 Chr.X: 72464913-72464913 on Build GRCh38
rs782753561 Chr.X: 72464664-72464664 on Build GRCh38
rs1005332612 Chr.X: 72464762-72464762 on Build GRCh38
rs782585859 Chr.X: 72464514-72464514 on Build GRCh38
rs782561917 Chr.X: 72464772-72464772 on Build GRCh38
rs886041838 Chr.X: 72464682-72464682 on Build GRCh38
rs782467454 Chr.X: 72464590-72464590 on Build GRCh38
rs782642759 Chr.X: 72464781-72464781 on Build GRCh38
rs797045613 Chr.X: 72464699-72464699 on Build GRCh38
rs782500602 Chr.X: 72464777-72464777 on Build GRCh38
rs782085368 Chr.X: 72464647-72464647 on Build GRCh38
rs781936429 Chr.X: 72464601-72464601 on Build GRCh38
rs1029588104 Chr.X: 72464895-72464895 on Build GRCh38
rs368682006 Chr.X: 72464713-72464713 on Build GRCh38
rs1015742979 Chr.X: 72464807-72464807 on Build GRCh38
rs781944571 Chr.X: 72464580-72464580 on Build GRCh38
rs952480301 Chr.X: 72464583-72464583 on Build GRCh38
rs1057516037 Chr.X: 72464626-72464630 on Build GRCh38
rs886039706 Chr.X: 72464562-72464562 on Build GRCh38
rs782294776 Chr.X: 72464735-72464735 on Build GRCh38
rs781878660 Chr.X: 72464635-72464635 on Build GRCh38
rs986143230 Chr.X: 72464954-72464954 on Build GRCh38
rs782243986 Chr.X: 72464547-72464547 on Build GRCh38
rs782412149 Chr.X: 72464527-72464527 on Build GRCh38

View Full Product Details