Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11668561-11668932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO6
Entrez Gene ID
26270
Gene Name
F-box protein 6
Gene Aliases
FBG2, FBS2, FBX6, Fbx6b
UniGene
Hs.464419
Gene Chromosome Location
Chr.1: 11664093-11674354 on Build GRCh38
Transcripts
NM_018438.5 XM_005263448.4 XM_005263449.4 XM_005263451.4 XM_006710571.3 XM_011541232.2 XM_017001007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161020 c.236A>G p.Y79C Chr.1: 11668894-11668894 on Build GRCh38
4684028 c.77C>T p.T26M Chr.1: 11668735-11668735 on Build GRCh38
2178568 c.151A>G p.M51V Chr.1: 11668809-11668809 on Build GRCh38
6662376 c.245G>A p.R82Q Chr.1: 11668903-11668903 on Build GRCh38
423557 c.179G>C p.R60P Chr.1: 11668837-11668837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546495045 Chr.1: 11668722-11668722 on Build GRCh38
rs775080648 Chr.1: 11668610-11668610 on Build GRCh38
rs370014977 Chr.1: 11668700-11668700 on Build GRCh38
rs773333033 Chr.1: 11668868-11668868 on Build GRCh38
rs756477067 Chr.1: 11668899-11668899 on Build GRCh38
rs138673974 Chr.1: 11668863-11668863 on Build GRCh38
rs771144830 Chr.1: 11668744-11668744 on Build GRCh38
rs774560160 Chr.1: 11668860-11668860 on Build GRCh38
rs751284987 Chr.1: 11668898-11668898 on Build GRCh38
rs771392177 Chr.1: 11668620-11668620 on Build GRCh38
rs571189937 Chr.1: 11668792-11668792 on Build GRCh38
rs780752818 Chr.1: 11668754-11668756 on Build GRCh38
rs764456551 Chr.1: 11668695-11668695 on Build GRCh38
rs370915217 Chr.1: 11668617-11668617 on Build GRCh38
rs778389623 Chr.1: 11668739-11668739 on Build GRCh38
rs138203471 Chr.1: 11668809-11668809 on Build GRCh38
rs763323423 Chr.1: 11668638-11668638 on Build GRCh38
rs931912503 Chr.1: 11668684-11668684 on Build GRCh38
rs781380213 Chr.1: 11668830-11668830 on Build GRCh38
rs561292765 Chr.1: 11668749-11668749 on Build GRCh38
rs748939144 Chr.1: 11668668-11668668 on Build GRCh38
rs200427755 Chr.1: 11668639-11668639 on Build GRCh38
rs141311977 Chr.1: 11668771-11668771 on Build GRCh38
rs780663319 Chr.1: 11668847-11668847 on Build GRCh38
rs753804712 Chr.1: 11668796-11668796 on Build GRCh38
rs1034236120 Chr.1: 11668594-11668594 on Build GRCh38
rs771360200 Chr.1: 11668678-11668678 on Build GRCh38
rs769979789 Chr.1: 11668687-11668687 on Build GRCh38
rs925299038 Chr.1: 11668796-11668813 on Build GRCh38
rs756929790 Chr.1: 11668633-11668634 on Build GRCh38
rs780939980 Chr.1: 11668646-11668646 on Build GRCh38
rs143110123 Chr.1: 11668824-11668824 on Build GRCh38
rs779599264 Chr.1: 11668667-11668667 on Build GRCh38
rs200248672 Chr.1: 11668896-11668896 on Build GRCh38
rs763235388 Chr.1: 11668692-11668692 on Build GRCh38
rs376458493 Chr.1: 11668625-11668625 on Build GRCh38
rs777216948 Chr.1: 11668621-11668621 on Build GRCh38
rs752289753 Chr.1: 11668888-11668888 on Build GRCh38
rs766167101 Chr.1: 11668872-11668872 on Build GRCh38
rs747842623 Chr.1: 11668740-11668740 on Build GRCh38
rs545601482 Chr.1: 11668673-11668673 on Build GRCh38
rs3125818 Chr.1: 11668837-11668837 on Build GRCh38
rs1052156546 Chr.1: 11668701-11668701 on Build GRCh38
rs749814822 Chr.1: 11668854-11668854 on Build GRCh38
rs761472382 Chr.1: 11668706-11668706 on Build GRCh38
rs1048391078 Chr.1: 11668770-11668770 on Build GRCh38
rs755566731 Chr.1: 11668650-11668650 on Build GRCh38
rs544960248 Chr.1: 11668600-11668600 on Build GRCh38
rs765412801 Chr.1: 11668623-11668623 on Build GRCh38
rs754420841 Chr.1: 11668802-11668802 on Build GRCh38
rs1014110632 Chr.1: 11668754-11668754 on Build GRCh38
rs773104067 Chr.1: 11668788-11668788 on Build GRCh38
rs144734810 Chr.1: 11668736-11668736 on Build GRCh38
rs957794457 Chr.1: 11668632-11668632 on Build GRCh38
rs776937731 Chr.1: 11668683-11668683 on Build GRCh38
rs768341299 Chr.1: 11668669-11668669 on Build GRCh38
rs767346215 Chr.1: 11668709-11668709 on Build GRCh38
rs934235702 Chr.1: 11668660-11668660 on Build GRCh38
rs772417342 Chr.1: 11668612-11668612 on Build GRCh38
rs781662877 Chr.1: 11668745-11668745 on Build GRCh38
rs923719128 Chr.1: 11668630-11668630 on Build GRCh38
rs141454413 Chr.1: 11668790-11668790 on Build GRCh38
rs144548369 Chr.1: 11668878-11668878 on Build GRCh38
rs7513972 Chr.1: 11668618-11668618 on Build GRCh38
rs139073449 Chr.1: 11668735-11668735 on Build GRCh38
rs746354058 Chr.1: 11668746-11668746 on Build GRCh38
rs955636873 Chr.1: 11668829-11668829 on Build GRCh38
rs762156094 Chr.1: 11668776-11668776 on Build GRCh38
rs922914255 Chr.1: 11668655-11668655 on Build GRCh38
rs1028657502 Chr.1: 11668870-11668870 on Build GRCh38
rs150942470 Chr.1: 11668877-11668877 on Build GRCh38
rs756113051 Chr.1: 11668717-11668717 on Build GRCh38
rs189560074 Chr.1: 11668685-11668685 on Build GRCh38
rs748957192 Chr.1: 11668611-11668611 on Build GRCh38
rs373737269 Chr.1: 11668710-11668710 on Build GRCh38
rs768884718 Chr.1: 11668751-11668751 on Build GRCh38
rs778911625 Chr.1: 11668908-11668908 on Build GRCh38
rs749944680 Chr.1: 11668902-11668902 on Build GRCh38
rs764817566 Chr.1: 11668803-11668803 on Build GRCh38
rs775779660 Chr.1: 11668689-11668689 on Build GRCh38
rs146139993 Chr.1: 11668750-11668750 on Build GRCh38
rs143708112 Chr.1: 11668903-11668903 on Build GRCh38
rs764647702 Chr.1: 11668886-11668886 on Build GRCh38
rs772390592 Chr.1: 11668865-11668865 on Build GRCh38
rs750403840 Chr.1: 11668649-11668649 on Build GRCh38
rs759496313 Chr.1: 11668622-11668622 on Build GRCh38
rs756565373 Chr.1: 11668839-11668839 on Build GRCh38
rs747288004 Chr.1: 11668675-11668675 on Build GRCh38
rs768557507 Chr.1: 11668857-11668857 on Build GRCh38
rs752908010 Chr.1: 11668631-11668631 on Build GRCh38
rs183656181 Chr.1: 11668641-11668641 on Build GRCh38
rs368549622 Chr.1: 11668871-11668871 on Build GRCh38
rs7514198 Chr.1: 11668895-11668895 on Build GRCh38
rs767214127 Chr.1: 11668785-11668785 on Build GRCh38

View Full Product Details