The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278469.1 | NP_001265398.1 | 21 - 22 | 2971 | 147 |
NM_001278470.1 | NP_001265399.1 | 21 - 22 | 2971 | 147 | |
NM_001278471.1 | NP_001265400.1 | 21 - 22 | 2902 | 147 | |
NM_001278472.1 | NP_001265401.1 | 22 - 23 | 3049 | 147 | |
NM_015026.2 | NP_055841.2 | 21 - 22 | 2971 | 147 | |
XM_017019041.1 | XP_016874530.1 | 22 - 23 | 2974 | 147 | |
XM_017019042.1 | XP_016874531.1 | 22 - 23 | 2974 | 147 | |
XM_017019043.1 | XP_016874532.1 | 23 - 24 | 3048 | 147 | |
XM_017019044.1 | XP_016874533.1 | 23 - 24 | 3050 | 147 | |
XM_017019045.1 | XP_016874534.1 | 24 - 25 | 2718 | 147 | |
XM_017019046.1 | XP_016874535.1 | 18 - 19 | 2143 | 147 | |
GenBank mRNA | AB017814.1 | - | 21 - 22 | 2696 | 147 |
AB028963.3 | - | 22 - 23 | 2942 | 147 | |
AL833066.2 | - | 21 - 22 | 2966 | 147 | |
AL834320.1 | - | 15 - 16 | 1929 | 147 | |
BC136621.1 | - | 21 - 22 | 2940 | 147 | |
BC141817.1 | - | 22 - 23 | 2942 | 147 | |
BC142710.1 | - | 22 - 23 | 2942 | 147 | |
BC144309.1 | - | 21 - 22 | 2871 | 147 | |
BC151241.1 | - | 22 - 23 | 2965 | 147 | |
BX537415.1 | - | 21 - 22 | 2969 | 147 |