Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GRIK1-AS1
- Entrez Gene ID
-
642976
- Gene Name
- GRIK1 antisense RNA 1
- Gene Aliases
-
C21orf9, GRIK1-AS, GRIK1AS, NCRNA00110
- Chromosome Location
-
Chr.21: 29748175 - 29764006 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY077697.1 |
- |
2 - 3 |
389 |
112 |
View Full Product Details