Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
Chromosome Location
Chr.15: 42723557 - 42737219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138477.2 NP_612486.2 14 - 15 2294 75
XM_005254176.4 XP_005254233.1 14 - 15 2209 75
XM_011521270.2 XP_011519572.1 14 - 15 2234 75
XM_011521271.2 XP_011519573.1 14 - 15 2231 75
XM_011521274.2 XP_011519576.1 11 - 12 1280 75
GenBank mRNA BC001092.2 - 6 - 7 749 75
BC008333.1 - 6 - 7 749 75
BC008334.1 - 6 - 7 749 75
BC052568.1 - 14 - 15 2182 75
BC066640.1 - 14 - 15 2182 75

View Full Product Details