The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008938.3 | NP_001008938.1 | 5 - 6 | 758 | 86 |
NM_014756.3 | NP_055571.2 | 5 - 6 | 758 | 86 | |
GenBank mRNA | BC070136.1 | - | 5 - 6 | 728 | 86 |
BC111043.1 | - | 5 - 6 | 710 | 86 | |
BC120869.1 | - | 4 - 5 | 652 | 86 | |
BC120870.1 | - | 4 - 5 | 650 | 86 | |
D43948.1 | - | 4 - 5 | 679 | 86 | |
X92474.1 | - | 4 - 5 | 658 | 86 |