The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015245.2 | NP_056060.2 | 21 - 22 | 3273 | 66 |
XM_005248964.3 | XP_005249021.1 | 22 - 23 | 3324 | 66 | |
XM_006715036.2 | XP_006715099.1 | 22 - 23 | 3327 | 66 | |
XM_011514431.2 | XP_011512733.1 | 22 - 23 | 3327 | 66 | |
XM_011514432.2 | XP_011512734.1 | 22 - 23 | 3324 | 66 | |
XM_011514433.1 | XP_011512735.1 | 22 - 23 | 3327 | 66 | |
XM_011514434.2 | XP_011512736.1 | 21 - 22 | 3264 | 66 | |
XM_011514435.2 | XP_011512737.1 | 21 - 22 | 3261 | 66 | |
XM_011514436.2 | XP_011512738.1 | 22 - 23 | 6896 | 66 | |
XM_011514437.2 | XP_011512739.1 | 12 - 13 | 3152 | 66 | |
XM_017010593.1 | XP_016866082.1 | 12 - 13 | 1750 | 66 | |
GenBank mRNA | BC031934.1 | - | 23 - 24 | 2939 | 66 |
BC049213.1 | - | 10 - 11 | 1116 | 66 | |
BC132832.1 | - | 21 - 22 | 3210 | 66 | |
BQ216321.1 | - | 6 - 7 | 656 | 66 | |
BX094969.1 | - | 4 - 5 | 402 | 66 | |
D86982.1 | - | 21 - 22 | 3270 | 66 |