The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014607.3 | NP_055422.1 | 1 - 2 | 387 | 60 |
GenBank mRNA | AK292829.1 | - | 1 - 2 | 80 | 60 |
AK304096.1 | - | 1 - 2 | 151 | 60 | |
AK311058.1 | - | 1 - 2 | 380 | 60 | |
BC020806.1 | - | 1 - 2 | 138 | 60 | |
BC035594.2 | - | 2 - 3 | 219 | 60 | |
BP247627.1 | - | 1 - 2 | 387 | 60 | |
BX647216.1 | - | 1 - 2 | 182 | 60 | |
BX647662.1 | - | 1 - 2 | 179 | 60 | |
D87684.1 | - | 1 - 2 | 140 | 60 | |
EF199840.1 | - | 1 - 2 | 204 | 60 |