Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN4
Entrez Gene ID
23190
Gene Name
UBX domain protein 4
Gene Aliases
UBXD2, UBXDC1, erasin
Chromosome Location
Chr.2: 135741619 - 135785063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014607.3 NP_055422.1 9 - 10 1266 81
GenBank mRNA AK026971.1 - 9 - 10 889 81
AK095402.1 - 5 - 6 654 81
AK292829.1 - 9 - 10 959 81
AK304096.1 - 9 - 10 976 81
BC035594.2 - 10 - 11 1098 81
BX647662.1 - 9 - 10 1058 81
D87684.1 - 9 - 10 1019 81
EF199840.1 - 9 - 10 1083 81

View Full Product Details