Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32666250-32666759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DQB1
Entrez Gene ID
3119
Gene Name
major histocompatibility complex, class II, DQ beta 1
Gene Aliases
CELIAC1, HLA-DQB, HLA-DRB1, IDDM1
UniGene
Hs.409934
Gene Chromosome Location
Chr.6: 32659464-32666689 on Build GRCh38
Transcripts
NM_001243961.1 NM_001243962.1 NM_002123.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3747678 c.35A>G p.D12G Chr.6: 32666573-32666573 on Build GRCh38
3830230 c.12G>A p.K4K Chr.6: 32666596-32666596 on Build GRCh38
451273 c.67G>T p.A23S Chr.6: 32666541-32666541 on Build GRCh38
6394024 c.103T>C p.S35P Chr.6: 32666505-32666505 on Build GRCh38
4161026 c.54C>G p.V18V Chr.6: 32666554-32666554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28746841 Chr.6: 32666317-32666317 on Build GRCh38
rs281860864 Chr.6: 32666477-32666477 on Build GRCh38
rs28746844 Chr.6: 32666362-32666362 on Build GRCh38
rs9274527 Chr.6: 32666713-32666713 on Build GRCh38
rs1130366 Chr.6: 32666523-32666523 on Build GRCh38
rs767634513 Chr.6: 32666565-32666565 on Build GRCh38
rs281861030 Chr.6: 32666298-32666298 on Build GRCh38
rs41315791 Chr.6: 32666393-32666393 on Build GRCh38
rs281860852 Chr.6: 32666491-32666491 on Build GRCh38
rs281860893 Chr.6: 32666446-32666446 on Build GRCh38
rs766997858 Chr.6: 32666520-32666521 on Build GRCh38
rs281861036 Chr.6: 32666291-32666291 on Build GRCh38
rs281861003 Chr.6: 32666329-32666329 on Build GRCh38
rs281860988 Chr.6: 32666344-32666344 on Build GRCh38
rs1029267040 Chr.6: 32666685-32666685 on Build GRCh38
rs281860846 Chr.6: 32666490-32666498 on Build GRCh38
rs281860941 Chr.6: 32666394-32666394 on Build GRCh38
rs281861047 Chr.6: 32666276-32666276 on Build GRCh38
rs3210146 Chr.6: 32666670-32666670 on Build GRCh38
rs772660704 Chr.6: 32666548-32666548 on Build GRCh38
rs281860970 Chr.6: 32666363-32666363 on Build GRCh38
rs281860951 Chr.6: 32666383-32666383 on Build GRCh38
rs281860968 Chr.6: 32666365-32666365 on Build GRCh38
rs281860967 Chr.6: 32666366-32666366 on Build GRCh38
rs281861042 Chr.6: 32666283-32666283 on Build GRCh38
rs66957193 Chr.6: 32666486-32666486 on Build GRCh38
rs138537247 Chr.6: 32666729-32666730 on Build GRCh38
rs281860871 Chr.6: 32666470-32666470 on Build GRCh38
rs281860872 Chr.6: 32666469-32666469 on Build GRCh38
rs281861014 Chr.6: 32666318-32666318 on Build GRCh38
rs281861023 Chr.6: 32666308-32666308 on Build GRCh38
rs281861004 Chr.6: 32666328-32666328 on Build GRCh38
rs281861007 Chr.6: 32666325-32666325 on Build GRCh38
rs28746845 Chr.6: 32666373-32666373 on Build GRCh38
rs281860913 Chr.6: 32666423-32666423 on Build GRCh38
rs281860932 Chr.6: 32666402-32666402 on Build GRCh38
rs28746851 Chr.6: 32666729-32666729 on Build GRCh38
rs2854260 Chr.6: 32666297-32666297 on Build GRCh38
rs770457774 Chr.6: 32666614-32666614 on Build GRCh38
rs281860898 Chr.6: 32666440-32666440 on Build GRCh38
rs28746846 Chr.6: 32666484-32666484 on Build GRCh38
rs281860897 Chr.6: 32666441-32666441 on Build GRCh38
rs200030215 Chr.6: 32666432-32666432 on Build GRCh38
rs28746840 Chr.6: 32666314-32666314 on Build GRCh38
rs281860851 Chr.6: 32666492-32666492 on Build GRCh38
rs796198016 Chr.6: 32666375-32666377 on Build GRCh38
rs281860989 Chr.6: 32666343-32666343 on Build GRCh38
rs281861035 Chr.6: 32666292-32666292 on Build GRCh38
rs281861010 Chr.6: 32666322-32666322 on Build GRCh38
rs281861029 Chr.6: 32666299-32666299 on Build GRCh38
rs281860990 Chr.6: 32666342-32666342 on Build GRCh38
rs281861013 Chr.6: 32666319-32666319 on Build GRCh38
rs28746847 Chr.6: 32666485-32666485 on Build GRCh38
rs281861012 Chr.6: 32666320-32666320 on Build GRCh38
rs386699587 Chr.6: 32666360-32666364 on Build GRCh38
rs281860927 Chr.6: 32666408-32666408 on Build GRCh38
rs796429679 Chr.6: 32666326-32666326 on Build GRCh38
rs9274513 Chr.6: 32666431-32666431 on Build GRCh38
rs28746843 Chr.6: 32666360-32666360 on Build GRCh38
rs281861041 Chr.6: 32666284-32666284 on Build GRCh38
rs753527747 Chr.6: 32666525-32666526 on Build GRCh38
rs281861008 Chr.6: 32666324-32666324 on Build GRCh38
rs1049056 Chr.6: 32666592-32666592 on Build GRCh38
rs281860892 Chr.6: 32666447-32666447 on Build GRCh38
rs281860891 Chr.6: 32666448-32666448 on Build GRCh38
rs281860888 Chr.6: 32666451-32666451 on Build GRCh38
rs281861009 Chr.6: 32666323-32666323 on Build GRCh38
rs758753379 Chr.6: 32666609-32666611 on Build GRCh38
rs281861022 Chr.6: 32666309-32666309 on Build GRCh38
rs281860953 Chr.6: 32666381-32666381 on Build GRCh38
rs3189152 Chr.6: 32666564-32666564 on Build GRCh38
rs1049061 Chr.6: 32666526-32666526 on Build GRCh38
rs281861045 Chr.6: 32666278-32666278 on Build GRCh38
rs281860929 Chr.6: 32666405-32666405 on Build GRCh38
rs746435665 Chr.6: 32666612-32666612 on Build GRCh38
rs9274511 Chr.6: 32666412-32666412 on Build GRCh38
rs200162904 Chr.6: 32666521-32666521 on Build GRCh38
rs281860934 Chr.6: 32666400-32666400 on Build GRCh38
rs368832944 Chr.6: 32666547-32666547 on Build GRCh38
rs281860933 Chr.6: 32666401-32666401 on Build GRCh38
rs281860938 Chr.6: 32666396-32666396 on Build GRCh38
rs281860972 Chr.6: 32666361-32666361 on Build GRCh38
rs9274512 Chr.6: 32666415-32666415 on Build GRCh38
rs281860876 Chr.6: 32666464-32666464 on Build GRCh38
rs281861016 Chr.6: 32666316-32666316 on Build GRCh38
rs281860877 Chr.6: 32666463-32666463 on Build GRCh38
rs12722106 Chr.6: 32666505-32666505 on Build GRCh38
rs281861024 Chr.6: 32666306-32666306 on Build GRCh38
rs281860870 Chr.6: 32666471-32666471 on Build GRCh38
rs281860899 Chr.6: 32666439-32666439 on Build GRCh38
rs66957194 Chr.6: 32666486-32666486 on Build GRCh38
rs766158480 Chr.6: 32666507-32666507 on Build GRCh38
rs756234491 Chr.6: 32666572-32666572 on Build GRCh38
rs281860944 Chr.6: 32666391-32666391 on Build GRCh38
rs281860943 Chr.6: 32666392-32666392 on Build GRCh38
rs796119008 Chr.6: 32666486-32666487 on Build GRCh38
rs9274504 Chr.6: 32666334-32666334 on Build GRCh38
rs281860823 Chr.6: 32666720-32666720 on Build GRCh38
rs281860935 Chr.6: 32666399-32666399 on Build GRCh38
rs3891175 Chr.6: 32666690-32666690 on Build GRCh38
rs117403290 Chr.6: 32666699-32666699 on Build GRCh38
rs386699588 Chr.6: 32666373-32666375 on Build GRCh38
rs41316196 Chr.6: 32666387-32666387 on Build GRCh38
rs778891121 Chr.6: 32666527-32666528 on Build GRCh38
rs281860937 Chr.6: 32666397-32666397 on Build GRCh38
rs281860945 Chr.6: 32666390-32666390 on Build GRCh38
rs281860850 Chr.6: 32666493-32666493 on Build GRCh38
rs281860886 Chr.6: 32666453-32666453 on Build GRCh38
rs281860885 Chr.6: 32666454-32666454 on Build GRCh38
rs281861038 Chr.6: 32666288-32666288 on Build GRCh38
rs281860954 Chr.6: 32666380-32666380 on Build GRCh38
rs1049059 Chr.6: 32666536-32666536 on Build GRCh38
rs281860919 Chr.6: 32666417-32666417 on Build GRCh38
rs281860918 Chr.6: 32666418-32666418 on Build GRCh38
rs28746842 Chr.6: 32666321-32666321 on Build GRCh38
rs34236112 Chr.6: 32666617-32666617 on Build GRCh38
rs281860920 Chr.6: 32666416-32666416 on Build GRCh38
rs281860974 Chr.6: 32666359-32666359 on Build GRCh38
rs9274514 Chr.6: 32666466-32666466 on Build GRCh38
rs1130373 Chr.6: 32666524-32666524 on Build GRCh38
rs769981529 Chr.6: 32666531-32666531 on Build GRCh38
rs281860996 Chr.6: 32666336-32666336 on Build GRCh38
rs574506463 Chr.6: 32666527-32666527 on Build GRCh38
rs281860931 Chr.6: 32666403-32666403 on Build GRCh38
rs281860979 Chr.6: 32666354-32666354 on Build GRCh38
rs9274503 Chr.6: 32666327-32666327 on Build GRCh38
rs761518101 Chr.6: 32666515-32666516 on Build GRCh38
rs281860955 Chr.6: 32666379-32666379 on Build GRCh38
rs281860980 Chr.6: 32666353-32666353 on Build GRCh38
rs281860956 Chr.6: 32666378-32666378 on Build GRCh38
rs281860858 Chr.6: 32666483-32666483 on Build GRCh38
rs281860900 Chr.6: 32666438-32666438 on Build GRCh38
rs775216584 Chr.6: 32666625-32666625 on Build GRCh38
rs281860869 Chr.6: 32666472-32666472 on Build GRCh38
rs281860873 Chr.6: 32666468-32666468 on Build GRCh38
rs281860930 Chr.6: 32666404-32666404 on Build GRCh38
rs281860875 Chr.6: 32666465-32666465 on Build GRCh38
rs9274510 Chr.6: 32666407-32666407 on Build GRCh38
rs281860946 Chr.6: 32666389-32666389 on Build GRCh38
rs9274525 Chr.6: 32666705-32666705 on Build GRCh38
rs281861037 Chr.6: 32666289-32666289 on Build GRCh38
rs766611168 Chr.6: 32666653-32666653 on Build GRCh38
rs281861033 Chr.6: 32666294-32666294 on Build GRCh38
rs1049072 Chr.6: 32666578-32666578 on Build GRCh38
rs72844397 Chr.6: 32666296-32666296 on Build GRCh38
rs750530800 Chr.6: 32666566-32666566 on Build GRCh38
rs281860849 Chr.6: 32666494-32666494 on Build GRCh38
rs71542466 Chr.6: 32666728-32666728 on Build GRCh38
rs281860822 Chr.6: 32666726-32666726 on Build GRCh38
rs281860992 Chr.6: 32666340-32666340 on Build GRCh38
rs1049053 Chr.6: 32666628-32666628 on Build GRCh38
rs34297496 Chr.6: 32666452-32666452 on Build GRCh38
rs68033958 Chr.6: 32666449-32666449 on Build GRCh38
rs281861044 Chr.6: 32666279-32666279 on Build GRCh38
rs937330135 Chr.6: 32666595-32666595 on Build GRCh38
rs281860901 Chr.6: 32666437-32666437 on Build GRCh38
rs281860914 Chr.6: 32666422-32666422 on Build GRCh38
rs1049057 Chr.6: 32666573-32666573 on Build GRCh38
rs772511655 Chr.6: 32666603-32666603 on Build GRCh38
rs766371114 Chr.6: 32666524-32666525 on Build GRCh38
rs281860859 Chr.6: 32666482-32666482 on Build GRCh38
rs759288400 Chr.6: 32666558-32666558 on Build GRCh38
rs281860855 Chr.6: 32666488-32666488 on Build GRCh38
rs796566642 Chr.6: 32666326-32666327 on Build GRCh38
rs281860936 Chr.6: 32666398-32666398 on Build GRCh38
rs281860908 Chr.6: 32666430-32666430 on Build GRCh38
rs281860975 Chr.6: 32666358-32666358 on Build GRCh38
rs281860976 Chr.6: 32666357-32666357 on Build GRCh38
rs281860939 Chr.6: 32666396-32666396 on Build GRCh38
rs9274508 Chr.6: 32666377-32666377 on Build GRCh38
rs68082907 Chr.6: 32666341-32666341 on Build GRCh38
rs796668956 Chr.6: 32666523-32666525 on Build GRCh38
rs281861021 Chr.6: 32666311-32666311 on Build GRCh38
rs281861020 Chr.6: 32666312-32666312 on Build GRCh38
rs748378081 Chr.6: 32666596-32666596 on Build GRCh38
rs36222416 Chr.6: 32666562-32666562 on Build GRCh38
rs9274505 Chr.6: 32666349-32666349 on Build GRCh38
rs749609676 Chr.6: 32666537-32666537 on Build GRCh38
rs281860993 Chr.6: 32666339-32666339 on Build GRCh38
rs1049055 Chr.6: 32666610-32666610 on Build GRCh38
rs67851506 Chr.6: 32666525-32666526 on Build GRCh38
rs71789649 Chr.6: 32666626-32666628 on Build GRCh38
rs281861027 Chr.6: 32666302-32666302 on Build GRCh38
rs28746839 Chr.6: 32666313-32666313 on Build GRCh38
rs551381124 Chr.6: 32666619-32666619 on Build GRCh38
rs1049062 Chr.6: 32666525-32666525 on Build GRCh38
rs281860896 Chr.6: 32666442-32666442 on Build GRCh38
rs281861039 Chr.6: 32666286-32666286 on Build GRCh38
rs281860867 Chr.6: 32666474-32666474 on Build GRCh38
rs281860866 Chr.6: 32666475-32666475 on Build GRCh38
rs754079233 Chr.6: 32666659-32666659 on Build GRCh38
rs281860940 Chr.6: 32666395-32666395 on Build GRCh38
rs907953592 Chr.6: 32666577-32666577 on Build GRCh38
rs281860884 Chr.6: 32666455-32666455 on Build GRCh38
rs281860923 Chr.6: 32666413-32666413 on Build GRCh38
rs9274522 Chr.6: 32666596-32666596 on Build GRCh38
rs72844399 Chr.6: 32666487-32666487 on Build GRCh38
rs281861017 Chr.6: 32666315-32666315 on Build GRCh38
rs749944694 Chr.6: 32666522-32666523 on Build GRCh38
rs281861025 Chr.6: 32666305-32666305 on Build GRCh38
rs281860915 Chr.6: 32666421-32666421 on Build GRCh38
rs281860947 Chr.6: 32666388-32666388 on Build GRCh38
rs281860902 Chr.6: 32666436-32666436 on Build GRCh38
rs67955939 Chr.6: 32666411-32666411 on Build GRCh38
rs281860917 Chr.6: 32666419-32666419 on Build GRCh38
rs66662713 Chr.6: 32666427-32666427 on Build GRCh38
rs281860916 Chr.6: 32666420-32666420 on Build GRCh38
rs9274529 Chr.6: 32666737-32666737 on Build GRCh38
rs281860959 Chr.6: 32666374-32666374 on Build GRCh38
rs281860964 Chr.6: 32666369-32666369 on Build GRCh38
rs281860963 Chr.6: 32666370-32666370 on Build GRCh38
rs281860862 Chr.6: 32666479-32666479 on Build GRCh38
rs754778659 Chr.6: 32666525-32666525 on Build GRCh38
rs281860978 Chr.6: 32666355-32666355 on Build GRCh38
rs41316200 Chr.6: 32666406-32666406 on Build GRCh38
rs281860848 Chr.6: 32666495-32666495 on Build GRCh38
rs281861026 Chr.6: 32666303-32666303 on Build GRCh38
rs28746838 Chr.6: 32666280-32666280 on Build GRCh38
rs201754614 Chr.6: 32666484-32666484 on Build GRCh38
rs281860994 Chr.6: 32666338-32666338 on Build GRCh38
rs1049060 Chr.6: 32666529-32666529 on Build GRCh38
rs281860909 Chr.6: 32666428-32666428 on Build GRCh38
rs281860997 Chr.6: 32666335-32666335 on Build GRCh38
rs281860999 Chr.6: 32666333-32666333 on Build GRCh38
rs281860922 Chr.6: 32666414-32666414 on Build GRCh38
rs281861000 Chr.6: 32666332-32666332 on Build GRCh38
rs200464814 Chr.6: 32666326-32666326 on Build GRCh38
rs281860860 Chr.6: 32666481-32666481 on Build GRCh38
rs281860861 Chr.6: 32666480-32666480 on Build GRCh38
rs281860895 Chr.6: 32666444-32666444 on Build GRCh38
rs281860854 Chr.6: 32666490-32666490 on Build GRCh38
rs281861040 Chr.6: 32666285-32666285 on Build GRCh38
rs754779441 Chr.6: 32666519-32666519 on Build GRCh38
rs281860949 Chr.6: 32666386-32666386 on Build GRCh38
rs281861034 Chr.6: 32666293-32666293 on Build GRCh38
rs780795041 Chr.6: 32666512-32666512 on Build GRCh38
rs281860868 Chr.6: 32666473-32666473 on Build GRCh38
rs9274506 Chr.6: 32666364-32666364 on Build GRCh38
rs281860883 Chr.6: 32666456-32666456 on Build GRCh38
rs920618234 Chr.6: 32666593-32666593 on Build GRCh38
rs281860878 Chr.6: 32666462-32666462 on Build GRCh38
rs281860995 Chr.6: 32666337-32666337 on Build GRCh38
rs3891176 Chr.6: 32666541-32666541 on Build GRCh38
rs281860984 Chr.6: 32666348-32666348 on Build GRCh38
rs750071545 Chr.6: 32666459-32666459 on Build GRCh38
rs281860982 Chr.6: 32666350-32666350 on Build GRCh38
rs281860981 Chr.6: 32666352-32666352 on Build GRCh38
rs281860985 Chr.6: 32666347-32666347 on Build GRCh38
rs281860986 Chr.6: 32666346-32666346 on Build GRCh38
rs878922085 Chr.6: 32666642-32666642 on Build GRCh38
rs281860903 Chr.6: 32666435-32666435 on Build GRCh38
rs281860880 Chr.6: 32666460-32666460 on Build GRCh38
rs281860879 Chr.6: 32666461-32666461 on Build GRCh38
rs9274509 Chr.6: 32666393-32666393 on Build GRCh38
rs281860911 Chr.6: 32666425-32666425 on Build GRCh38
rs281860961 Chr.6: 32666372-32666372 on Build GRCh38
rs281860977 Chr.6: 32666356-32666356 on Build GRCh38
rs281860847 Chr.6: 32666497-32666497 on Build GRCh38
rs281860965 Chr.6: 32666368-32666368 on Build GRCh38
rs281860910 Chr.6: 32666426-32666426 on Build GRCh38
rs281860925 Chr.6: 32666410-32666410 on Build GRCh38
rs183607390 Chr.6: 32666450-32666450 on Build GRCh38
rs750064618 Chr.6: 32666502-32666502 on Build GRCh38
rs117000147 Chr.6: 32666281-32666281 on Build GRCh38
rs368359746 Chr.6: 32666307-32666307 on Build GRCh38
rs372512148 Chr.6: 32666539-32666539 on Build GRCh38
rs281860894 Chr.6: 32666445-32666445 on Build GRCh38
rs281860906 Chr.6: 32666432-32666432 on Build GRCh38
rs9274528 Chr.6: 32666731-32666731 on Build GRCh38
rs281860863 Chr.6: 32666478-32666478 on Build GRCh38
rs281860987 Chr.6: 32666345-32666345 on Build GRCh38
rs1049070 Chr.6: 32666648-32666648 on Build GRCh38
rs1049058 Chr.6: 32666554-32666554 on Build GRCh38
rs281860865 Chr.6: 32666476-32666476 on Build GRCh38
rs281860882 Chr.6: 32666457-32666457 on Build GRCh38
rs753690273 Chr.6: 32666515-32666515 on Build GRCh38
rs281860881 Chr.6: 32666458-32666458 on Build GRCh38
rs369709024 Chr.6: 32666563-32666563 on Build GRCh38
rs281860853 Chr.6: 32666490-32666490 on Build GRCh38
rs281860952 Chr.6: 32666382-32666382 on Build GRCh38
rs575475309 Chr.6: 32666540-32666540 on Build GRCh38
rs386406640 Chr.6: 32666310-32666310 on Build GRCh38
rs281861001 Chr.6: 32666331-32666331 on Build GRCh38
rs281861002 Chr.6: 32666330-32666330 on Build GRCh38
rs281860912 Chr.6: 32666424-32666424 on Build GRCh38
rs9274526 Chr.6: 32666708-32666708 on Build GRCh38
rs281860904 Chr.6: 32666434-32666434 on Build GRCh38
rs111774873 Chr.6: 32666376-32666376 on Build GRCh38
rs67147537 Chr.6: 32666649-32666649 on Build GRCh38
rs281860905 Chr.6: 32666433-32666433 on Build GRCh38
rs281860926 Chr.6: 32666409-32666409 on Build GRCh38
rs281860966 Chr.6: 32666367-32666367 on Build GRCh38
rs281861028 Chr.6: 32666301-32666301 on Build GRCh38
rs281860950 Chr.6: 32666384-32666384 on Build GRCh38
rs9274507 Chr.6: 32666375-32666375 on Build GRCh38
rs371168889 Chr.6: 32666652-32666652 on Build GRCh38
rs281860962 Chr.6: 32666371-32666371 on Build GRCh38

View Full Product Details