The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017617.4 | NP_060087.3 | 22 - 23 | 3638 | 71 |
XM_011518717.2 | XP_011517019.2 | 19 - 20 | 2930 | 71 | |
GenBank mRNA | AB209873.1 | - | 14 - 15 | 2175 | 71 |
AF308602.1 | - | 22 - 23 | 3641 | 71 | |
M73980.1 | - | 22 - 23 | 3641 | 71 |