Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMNB2
Entrez Gene ID
84823
Gene Name
lamin B2
Gene Aliases
EPM9, LAMB2, LMN2
Chromosome Location
Chr.19: 2428165 - 2456968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032737.3 NP_116126.3 6 - 7 1014 73
GenBank mRNA BC006551.2 - 6 - 7 979 73
BT007441.1 - 6 - 7 919 73
KP137589.1 - 3 - 4 568 73
M94362.1 - 5 - 6 667 73

View Full Product Details