Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAZ3
Entrez Gene ID
57054
Gene Name
deleted in azoospermia 3
Gene Aliases
pDP1679
Chromosome Location
Chr.Y: 24763069 - 24813492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020364.3 NP_065097.2 17 - 18 1558 81
GenBank mRNA AF248481.1 - 19 - 20 1583 81
BC113005.1 - 17 - 18 1343 81

Gene Symbol
DAZ1
Entrez Gene ID
1617
Gene Name
deleted in azoospermia 1
Gene Aliases
DAZ, SPGY
Chromosome Location
Chr.Y: 23129355 - 23199117 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004081.5 NP_004072.3 26 - 27 2461 81
XM_011531482.2 XP_011529784.1 26 - 27 2499 81
XM_011531483.1 XP_011529785.1 20 - 21 1626 81
GenBank mRNA AF271087.1 - 14 - 15 1838 81
AF271088.1 - 18 - 19 1441 81
AK310133.1 - 2 - 3 232 81

Gene Symbol
DAZ2
Entrez Gene ID
57055
Gene Name
deleted in azoospermia 2
Gene Aliases
pDP1678
Chromosome Location
Chr.Y: 23219457 - 23291356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005785.2 NP_001005785.1 21 - 22 1823 81
NM_001005786.2 NP_001005786.2 14 - 15 1319 81
NM_020363.3 NP_065096.2 22 - 23 1895 81
GenBank mRNA AF248480.1 - 22 - 23 1807 81
AF414184.1 - 21 - 22 1805 81
AK302398.1 - 24 - 25 1962 81
AK302690.1 - 25 - 26 1964 81
BC113006.1 - 21 - 22 1631 81
U21663.1 - 14 - 15 1243 81

Gene Symbol
DAZ4
Entrez Gene ID
57135
Gene Name
deleted in azoospermia 4
Gene Aliases
pDP1680, pDP1681
Chromosome Location
Chr.Y: 24833807 - 24907040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005375.2 NP_001005375.1 21 - 22 1981 81
NM_020420.3 NP_065153.1 15 - 16 1414 81
XM_005262602.1 XP_005262659.1 27 - 28 2352 81
XM_011531509.2 XP_011529811.1 27 - 28 2439 81
XM_011531510.1 XP_011529812.1 21 - 22 1563 81
GenBank mRNA AF248483.1 - 15 - 16 1176 81
BC047480.1 - 15 - 16 1319 81
BC047617.1 - 15 - 16 1315 81

View Full Product Details