The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004095.3 | NP_004086.1 | 2 - 3 | 393 | 74 |
GenBank mRNA | AB044548.1 | - | 2 - 3 | 366 | 74 |
BC004459.1 | - | 2 - 3 | 385 | 74 | |
BC058073.1 | - | 2 - 3 | 333 | 74 |