Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86949901-86950444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD4
Entrez Gene ID
8322
Gene Name
frizzled class receptor 4
Gene Aliases
CD344, EVR1, FEVR, FZD4S, Fz-4, Fz4, FzE4, GPCR, hFz4
UniGene
Hs.591968
Gene Chromosome Location
Chr.11: 86945675-86955398 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

SNP Detail

SNP ID Chromosome Location
rs962484819 Chr.11: 86950193-86950193 on Build GRCh38
rs974410996 Chr.11: 86950280-86950280 on Build GRCh38
rs528457279 Chr.11: 86950258-86950258 on Build GRCh38
rs147951354 Chr.11: 86950022-86950022 on Build GRCh38
rs771583415 Chr.11: 86950255-86950255 on Build GRCh38
rs542063724 Chr.11: 86950334-86950334 on Build GRCh38
rs1025232427 Chr.11: 86950203-86950203 on Build GRCh38
rs954657716 Chr.11: 86950331-86950331 on Build GRCh38
rs553868043 Chr.11: 86950225-86950225 on Build GRCh38
rs898511085 Chr.11: 86950094-86950094 on Build GRCh38
rs972996837 Chr.11: 86950362-86950362 on Build GRCh38
rs757495000 Chr.11: 86950419-86950421 on Build GRCh38
rs766470922 Chr.11: 86950224-86950225 on Build GRCh38
rs1010042125 Chr.11: 86950063-86950063 on Build GRCh38
rs775197705 Chr.11: 86950363-86950363 on Build GRCh38
rs758765834 Chr.11: 86950024-86950024 on Build GRCh38
rs1013412045 Chr.11: 86950200-86950200 on Build GRCh38
rs1051737581 Chr.11: 86950005-86950005 on Build GRCh38
rs1003228169 Chr.11: 86949944-86949944 on Build GRCh38
rs551288994 Chr.11: 86950406-86950406 on Build GRCh38
rs920670091 Chr.11: 86950197-86950197 on Build GRCh38
rs891713127 Chr.11: 86950307-86950307 on Build GRCh38
rs995566875 Chr.11: 86950119-86950119 on Build GRCh38
rs988157771 Chr.11: 86950177-86950177 on Build GRCh38
rs966619488 Chr.11: 86950212-86950212 on Build GRCh38
rs932824383 Chr.11: 86949941-86949941 on Build GRCh38
rs141895567 Chr.11: 86950195-86950195 on Build GRCh38
rs573013571 Chr.11: 86950343-86950343 on Build GRCh38
rs904286116 Chr.11: 86950080-86950080 on Build GRCh38
rs1015189410 Chr.11: 86950192-86950192 on Build GRCh38
rs573091130 Chr.11: 86950326-86950326 on Build GRCh38
rs890087637 Chr.11: 86950407-86950407 on Build GRCh38
rs999413428 Chr.11: 86950237-86950237 on Build GRCh38
rs145679440 Chr.11: 86950378-86950378 on Build GRCh38
rs1021289329 Chr.11: 86950073-86950073 on Build GRCh38
rs752918590 Chr.11: 86950021-86950021 on Build GRCh38
rs1028644307 Chr.11: 86950385-86950385 on Build GRCh38
rs537438989 Chr.11: 86950004-86950004 on Build GRCh38
rs182907379 Chr.11: 86949982-86949982 on Build GRCh38
rs538226404 Chr.11: 86950092-86950092 on Build GRCh38
rs1057409354 Chr.11: 86950268-86950268 on Build GRCh38
rs779102119 Chr.11: 86950016-86950016 on Build GRCh38
rs61088092 Chr.11: 86950315-86950315 on Build GRCh38
rs188015808 Chr.11: 86950044-86950044 on Build GRCh38
rs891596602 Chr.11: 86950039-86950039 on Build GRCh38
rs954964899 Chr.11: 86950144-86950144 on Build GRCh38
rs940225405 Chr.11: 86950344-86950344 on Build GRCh38
rs80075834 Chr.11: 86950150-86950150 on Build GRCh38
rs1055195675 Chr.11: 86950126-86950126 on Build GRCh38
rs745378991 Chr.11: 86950259-86950259 on Build GRCh38

View Full Product Details