Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPPA2
Entrez Gene ID
151871
Gene Name
developmental pluripotency associated 2
Gene Aliases
CT100, ECAT15-2, PESCRG1
Chromosome Location
Chr.3: 109293788 - 109316553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138815.3 NP_620170.3 1 - 2 237 66
GenBank mRNA BC018070.1 - 1 - 2 115 66

View Full Product Details