Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYADM
Entrez Gene ID
91663
Gene Name
myeloid associated differentiation marker
Gene Aliases
SB135
Chromosome Location
Chr.19: 53865584 - 53876435 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020818.2 NP_001018654.1 1 - 2 450 58
NM_001020819.2 NP_001018655.1 2 - 3 230 58
NM_001020820.2 NP_001018656.1 2 - 3 347 58
NM_001020821.2 NP_001018657.1 2 - 3 295 58
NM_001290188.1 NP_001277117.1 2 - 3 411 58
NM_001290190.1 NP_001277119.1 2 - 3 202 58
NM_001290191.1 NP_001277120.1 2 - 3 154 58
NM_001290192.1 NP_001277121.1 2 - 3 171 58
NM_001290193.1 NP_001277122.1 2 - 3 232 58
NM_138373.4 NP_612382.1 2 - 3 166 58
GenBank mRNA AF087882.1 - 2 - 3 280 58
AK075276.1 - 2 - 3 148 58
AK093999.1 - 2 - 3 146 58
AK294400.1 - 2 - 3 148 58
AK316566.1 - 2 - 3 144 58
AX775819.1 - 2 - 3 146 58
AY037147.2 - 2 - 3 169 58
AY358582.1 - 2 - 3 130 58
BC013995.2 - 1 - 2 402 58
BC095412.1 - 2 - 3 148 58
BQ430033.1 - 2 - 3 232 58
BQ716724.1 - 2 - 3 140 58
BX393859.2 - 2 - 3 109 58
CN277531.1 - 2 - 3 172 58
CN284532.1 - 2 - 3 282 58
DA027466.1 - 2 - 3 215 58
DA472260.1 - 2 - 3 136 58
DA677314.1 - 2 - 3 202 58
DA948116.1 - 2 - 3 166 58
DC379591.1 - 2 - 3 411 58

View Full Product Details