Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL17A
Entrez Gene ID
51326
Gene Name
ADP ribosylation factor like GTPase 17A
Gene Aliases
ARF1P2, ARL17P1
Chromosome Location
Chr.17: 46500213 - 46579781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011524876.2 XP_011523178.1 4 - 5 715 96
XM_017024729.1 XP_016880218.1 4 - 5 715 96
GenBank mRNA BC000924.3 - 1 - 2 266 96

Gene Symbol
ARL17B
Entrez Gene ID
100506084
Gene Name
ADP ribosylation factor like GTPase 17B
Gene Aliases
ARL17, ARL17A
Chromosome Location
Chr.17: 46274184 - 46361872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011524163.2 XP_011522465.1 4 - 5 735 96

View Full Product Details