Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62597410-62597843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL3
Entrez Gene ID
27329
Gene Name
angiopoietin like 3
Gene Aliases
ANG-5, ANGPT5, ANL3, FHBL2
UniGene
Hs.209153
Gene Chromosome Location
Chr.1: 62597487-62606305 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
UniGene
Hs.744927
Gene Chromosome Location
Chr.1: 62454726-62688368 on Build GRCh38
Transcripts
NM_014495.3 NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_001272002.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_011542330.2 XM_017002639.1 XM_017002640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4929543 c.30T>A p.I10I Chr.1: 62597596-62597596 on Build GRCh38
3491638 c.85C>A p.L29I Chr.1: 62597651-62597651 on Build GRCh38
5053629 c.223C>A p.L75I Chr.1: 62597789-62597789 on Build GRCh38
363564 c.50_51CC>AA p.S17>? Chr.1: 62597616-62597617 on Build GRCh38
2238418 c.243delT p.Y83fs*17 Chr.1: 62597809-62597809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746347553 Chr.1: 62597635-62597642 on Build GRCh38
rs779312263 Chr.1: 62597604-62597604 on Build GRCh38
rs768802507 Chr.1: 62597558-62597558 on Build GRCh38
rs745885061 Chr.1: 62597530-62597530 on Build GRCh38
rs775299081 Chr.1: 62597782-62597782 on Build GRCh38
rs755058329 Chr.1: 62597763-62597763 on Build GRCh38
rs756096244 Chr.1: 62597677-62597677 on Build GRCh38
rs139687521 Chr.1: 62597717-62597717 on Build GRCh38
rs151064896 Chr.1: 62597715-62597715 on Build GRCh38
rs143189908 Chr.1: 62597642-62597642 on Build GRCh38
rs774569842 Chr.1: 62597698-62597698 on Build GRCh38
rs898614906 Chr.1: 62597531-62597531 on Build GRCh38
rs760521061 Chr.1: 62597576-62597576 on Build GRCh38
rs145578974 Chr.1: 62597783-62597783 on Build GRCh38
rs370895328 Chr.1: 62597749-62597749 on Build GRCh38
rs147500008 Chr.1: 62597692-62597692 on Build GRCh38
rs763781510 Chr.1: 62597738-62597738 on Build GRCh38
rs751955393 Chr.1: 62597636-62597636 on Build GRCh38
rs766843384 Chr.1: 62597819-62597819 on Build GRCh38
rs575552258 Chr.1: 62597725-62597725 on Build GRCh38
rs775056868 Chr.1: 62597731-62597731 on Build GRCh38
rs764827703 Chr.1: 62597617-62597617 on Build GRCh38
rs753899075 Chr.1: 62597758-62597758 on Build GRCh38
rs967302006 Chr.1: 62597505-62597505 on Build GRCh38
rs746743905 Chr.1: 62597700-62597700 on Build GRCh38
rs772332226 Chr.1: 62597766-62597766 on Build GRCh38
rs1032082501 Chr.1: 62597699-62597699 on Build GRCh38
rs751260529 Chr.1: 62597740-62597740 on Build GRCh38
rs781736614 Chr.1: 62597651-62597651 on Build GRCh38
rs748393501 Chr.1: 62597765-62597765 on Build GRCh38
rs752991282 Chr.1: 62597513-62597513 on Build GRCh38
rs952531091 Chr.1: 62597578-62597578 on Build GRCh38
rs780153441 Chr.1: 62597689-62597689 on Build GRCh38
rs12565895 Chr.1: 62597506-62597506 on Build GRCh38
rs759491919 Chr.1: 62597723-62597723 on Build GRCh38
rs776558413 Chr.1: 62597606-62597606 on Build GRCh38
rs369780744 Chr.1: 62597773-62597773 on Build GRCh38
rs199772471 Chr.1: 62597690-62597690 on Build GRCh38
rs768014539 Chr.1: 62597748-62597748 on Build GRCh38
rs765123937 Chr.1: 62597724-62597724 on Build GRCh38
rs182991271 Chr.1: 62597515-62597515 on Build GRCh38
rs746416387 Chr.1: 62597660-62597660 on Build GRCh38
rs1013469622 Chr.1: 62597619-62597619 on Build GRCh38
rs982453920 Chr.1: 62597611-62597611 on Build GRCh38
rs757596386 Chr.1: 62597522-62597522 on Build GRCh38
rs768313034 Chr.1: 62597789-62597789 on Build GRCh38
rs908723680 Chr.1: 62597490-62597490 on Build GRCh38
rs756641927 Chr.1: 62597757-62597757 on Build GRCh38
rs752195217 Chr.1: 62597629-62597629 on Build GRCh38
rs767345973 Chr.1: 62597801-62597801 on Build GRCh38
rs10449755 Chr.1: 62597553-62597553 on Build GRCh38
rs373926508 Chr.1: 62597817-62597817 on Build GRCh38
rs141150415 Chr.1: 62597809-62597809 on Build GRCh38
rs541408414 Chr.1: 62597529-62597529 on Build GRCh38
rs146749211 Chr.1: 62597754-62597754 on Build GRCh38
rs1011577448 Chr.1: 62597460-62597460 on Build GRCh38
rs546051366 Chr.1: 62597746-62597746 on Build GRCh38
rs533410817 Chr.1: 62597737-62597737 on Build GRCh38
rs398122986 Chr.1: 62597621-62597621 on Build GRCh38
rs746135169 Chr.1: 62597639-62597639 on Build GRCh38
rs763902658 Chr.1: 62597635-62597635 on Build GRCh38
rs766251086 Chr.1: 62597583-62597583 on Build GRCh38
rs1019431790 Chr.1: 62597535-62597535 on Build GRCh38
rs772988115 Chr.1: 62597570-62597570 on Build GRCh38
rs920003542 Chr.1: 62597815-62597815 on Build GRCh38
rs568241193 Chr.1: 62597492-62597492 on Build GRCh38
rs745714568 Chr.1: 62597774-62597774 on Build GRCh38
rs758931786 Chr.1: 62597616-62597616 on Build GRCh38
rs201052110 Chr.1: 62597804-62597804 on Build GRCh38
rs762424593 Chr.1: 62597734-62597734 on Build GRCh38
rs779071619 Chr.1: 62597764-62597764 on Build GRCh38
rs149086653 Chr.1: 62597794-62597794 on Build GRCh38
rs769753498 Chr.1: 62597778-62597778 on Build GRCh38
rs769920975 Chr.1: 62597537-62597537 on Build GRCh38
rs267606655 Chr.1: 62597616-62597617 on Build GRCh38
rs770747783 Chr.1: 62597709-62597709 on Build GRCh38
rs779437563 Chr.1: 62597633-62597634 on Build GRCh38
rs775532151 Chr.1: 62597538-62597538 on Build GRCh38

View Full Product Details