Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150218709-150219280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32E
Entrez Gene ID
81611
Gene Name
acidic nuclear phosphoprotein 32 family member E
Gene Aliases
LANP-L, LANPL
UniGene
Hs.656466
Gene Chromosome Location
Chr.1: 150218417-150236156 on Build GRCh38
Transcripts
NM_001136478.3 NM_001136479.2 NM_001280559.1 NM_001280560.1 NM_030920.4 XM_005245513.3 XM_005245514.3 XM_017002418.1

SNP Detail

SNP ID Chromosome Location
rs1039783083 Chr.1: 150218874-150218874 on Build GRCh38
rs587643148 Chr.1: 150219166-150219166 on Build GRCh38
rs782366466 Chr.1: 150219003-150219003 on Build GRCh38
rs904485540 Chr.1: 150218832-150218832 on Build GRCh38
rs587749829 Chr.1: 150219135-150219135 on Build GRCh38
rs1040091798 Chr.1: 150219133-150219133 on Build GRCh38
rs887064962 Chr.1: 150218933-150218933 on Build GRCh38
rs587695652 Chr.1: 150219071-150219071 on Build GRCh38
rs1043120327 Chr.1: 150218801-150218801 on Build GRCh38
rs1047322897 Chr.1: 150218929-150218929 on Build GRCh38
rs993047947 Chr.1: 150219016-150219016 on Build GRCh38
rs934062257 Chr.1: 150219207-150219207 on Build GRCh38
rs181404714 Chr.1: 150218914-150218914 on Build GRCh38
rs36050205 Chr.1: 150218973-150218973 on Build GRCh38
rs188989339 Chr.1: 150218837-150218837 on Build GRCh38
rs942770322 Chr.1: 150218857-150218857 on Build GRCh38
rs182816772 Chr.1: 150219231-150219231 on Build GRCh38
rs998052018 Chr.1: 150218918-150218918 on Build GRCh38
rs913390021 Chr.1: 150219021-150219021 on Build GRCh38
rs942654414 Chr.1: 150219084-150219084 on Build GRCh38
rs1005422246 Chr.1: 150218950-150218950 on Build GRCh38
rs186321795 Chr.1: 150219026-150219026 on Build GRCh38
rs114811641 Chr.1: 150219064-150219064 on Build GRCh38
rs1001920898 Chr.1: 150218997-150218997 on Build GRCh38
rs190688581 Chr.1: 150219202-150219202 on Build GRCh38
rs978974754 Chr.1: 150219027-150219027 on Build GRCh38
rs782238593 Chr.1: 150218967-150218967 on Build GRCh38
rs946275149 Chr.1: 150218798-150218798 on Build GRCh38
rs587753965 Chr.1: 150218761-150218761 on Build GRCh38
rs781955735 Chr.1: 150219008-150219008 on Build GRCh38
rs968989485 Chr.1: 150218974-150218974 on Build GRCh38

View Full Product Details