The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027159.1 | 2 - 3 | 498 | 75 | |
NR_027160.1 | 3 - 4 | 621 | 75 | ||
NR_027162.1 | 3 - 4 | 674 | 75 | ||
NR_027163.1 | 4 - 5 | 408 | 75 | ||
NR_027164.1 | 4 - 5 | 355 | 75 | ||
NR_027166.1 | 3 - 4 | 232 | 75 | ||
NR_027168.2 | 4 - 5 | 324 | 75 | ||
NR_027170.1 | 4 - 5 | 276 | 75 | ||
NR_027171.1 | 2 - 3 | 498 | 75 | ||
NR_027172.2 | 3 - 4 | 201 | 75 | ||
NR_027173.1 | 3 - 4 | 232 | 75 | ||
NR_027174.1 | 3 - 4 | 232 | 75 | ||
NR_027175.1 | 3 - 4 | 232 | 75 | ||
NR_027177.1 | 3 - 4 | 232 | 75 | ||
NR_027178.1 | 3 - 4 | 232 | 75 | ||
NR_027179.1 | 3 - 4 | 232 | 75 | ||
NR_027667.1 | 3 - 4 | 542 | 75 | ||
NR_045021.1 | 3 - 4 | 211 | 75 | ||
NR_045022.1 | 4 - 5 | 350 | 75 | ||
NR_045024.1 | 2 - 3 | 498 | 75 | ||
NR_045025.1 | 2 - 3 | 144 | 75 | ||
NR_045026.1 | 4 - 5 | 408 | 75 | ||
NR_045029.1 | 3 - 4 | 306 | 75 | ||
GenBank mRNA | AK098643.1 | - | 3 - 4 | 182 | 75 |
AK307982.1 | - | 3 - 4 | 624 | 75 | |
AK311247.1 | - | 3 - 4 | 788 | 75 | |
BC019833.1 | - | 2 - 3 | 158 | 75 | |
BC027986.1 | - | 3 - 4 | 492 | 75 | |
BC040159.1 | - | 3 - 4 | 471 | 75 | |
BQ070126.1 | - | 2 - 3 | 236 | 75 | |
BU506593.1 | - | - | 339 | 75 | |
BU599615.1 | - | 4 - 5 | 362 | 75 | |
BX538291.1 | - | 4 - 5 | 227 | 75 | |
CB150672.1 | - | 4 - 5 | 305 | 75 | |
CD050014.1 | - | 3 - 4 | 205 | 75 | |
CF126952.1 | - | 2 - 3 | 414 | 75 | |
CR989654.1 | - | 3 - 4 | 285 | 75 | |
DA131201.1 | - | 3 - 4 | 182 | 75 | |
DA193585.1 | - | 3 - 4 | 367 | 75 | |
DA383448.1 | - | 3 - 4 | 182 | 75 | |
DA440445.1 | - | 4 - 5 | 301 | 75 | |
DA485648.1 | - | 3 - 4 | 183 | 75 | |
DA494627.1 | - | 2 - 3 | 449 | 75 | |
DA793334.1 | - | 2 - 3 | 98 | 75 | |
DA802938.1 | - | 3 - 4 | 182 | 75 | |
DA857697.1 | - | 3 - 4 | 182 | 75 | |
DA867958.1 | - | 4 - 5 | 360 | 75 | |
DB258221.1 | - | 2 - 3 | 449 | 75 |