Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161505241-161505761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR2A
Entrez Gene ID
2212
Gene Name
Fc fragment of IgG receptor IIa
Gene Aliases
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
UniGene
Hs.352642
Gene Chromosome Location
Chr.1: 161505415-161524048 on Build GRCh38
Transcripts
NM_001136219.1 NM_021642.3 XM_011509287.2 XM_011509290.2 XM_011509291.1 XM_017000663.1 XM_017000664.1 XM_017000665.1 XM_017000666.1 XM_017000667.1 XM_017000668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252304 c.6T>C p.T2T Chr.1: 161505473-161505473 on Build GRCh38
6059017 c.39C>T p.P13P Chr.1: 161505506-161505506 on Build GRCh38
236107 c.8T>C p.M3T Chr.1: 161505475-161505475 on Build GRCh38
5006805 c.85+2T>G p.? Chr.1: 161505554-161505554 on Build GRCh38
898557 c.38C>T p.P13L Chr.1: 161505505-161505505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016815119 Chr.1: 161505690-161505690 on Build GRCh38
rs958303575 Chr.1: 161505301-161505301 on Build GRCh38
rs962725563 Chr.1: 161505729-161505729 on Build GRCh38
rs779537504 Chr.1: 161505580-161505580 on Build GRCh38
rs577732390 Chr.1: 161505491-161505491 on Build GRCh38
rs763426893 Chr.1: 161505525-161505525 on Build GRCh38
rs367608816 Chr.1: 161505346-161505346 on Build GRCh38
rs982920196 Chr.1: 161505704-161505704 on Build GRCh38
rs113343829 Chr.1: 161505583-161505583 on Build GRCh38
rs559534771 Chr.1: 161505563-161505563 on Build GRCh38
rs983978982 Chr.1: 161505347-161505347 on Build GRCh38
rs757837431 Chr.1: 161505577-161505577 on Build GRCh38
rs374230075 Chr.1: 161505601-161505601 on Build GRCh38
rs753552412 Chr.1: 161505555-161505555 on Build GRCh38
rs530369256 Chr.1: 161505678-161505678 on Build GRCh38
rs908507298 Chr.1: 161505472-161505472 on Build GRCh38
rs958571344 Chr.1: 161505326-161505326 on Build GRCh38
rs766483279 Chr.1: 161505528-161505528 on Build GRCh38
rs141594049 Chr.1: 161505497-161505497 on Build GRCh38
rs188798727 Chr.1: 161505594-161505594 on Build GRCh38
rs376631122 Chr.1: 161505496-161505496 on Build GRCh38
rs565005790 Chr.1: 161505605-161505605 on Build GRCh38
rs573934607 Chr.1: 161505537-161505537 on Build GRCh38
rs1021634755 Chr.1: 161505608-161505608 on Build GRCh38
rs752613385 Chr.1: 161505542-161505542 on Build GRCh38
rs563908162 Chr.1: 161505639-161505639 on Build GRCh38
rs530825369 Chr.1: 161505620-161505620 on Build GRCh38
rs908624156 Chr.1: 161505723-161505723 on Build GRCh38
rs200771460 Chr.1: 161505450-161505450 on Build GRCh38
rs909788587 Chr.1: 161505380-161505380 on Build GRCh38
rs983946172 Chr.1: 161505680-161505680 on Build GRCh38
rs575432232 Chr.1: 161505455-161505455 on Build GRCh38
rs773437367 Chr.1: 161505523-161505523 on Build GRCh38
rs757849128 Chr.1: 161505336-161505336 on Build GRCh38
rs921495802 Chr.1: 161505538-161505538 on Build GRCh38
rs770182244 Chr.1: 161505588-161505588 on Build GRCh38
rs902891805 Chr.1: 161505600-161505600 on Build GRCh38
rs111736397 Chr.1: 161505306-161505306 on Build GRCh38
rs774749129 Chr.1: 161505529-161505529 on Build GRCh38
rs745534149 Chr.1: 161505572-161505572 on Build GRCh38
rs1038672527 Chr.1: 161505527-161505527 on Build GRCh38
rs550517994 Chr.1: 161505629-161505629 on Build GRCh38
rs1032530335 Chr.1: 161505295-161505295 on Build GRCh38
rs919225165 Chr.1: 161505403-161505403 on Build GRCh38
rs1046445261 Chr.1: 161505438-161505438 on Build GRCh38
rs777432732 Chr.1: 161505479-161505479 on Build GRCh38
rs369444267 Chr.1: 161505452-161505452 on Build GRCh38
rs949420970 Chr.1: 161505422-161505422 on Build GRCh38
rs749170200 Chr.1: 161505481-161505481 on Build GRCh38
rs746538691 Chr.1: 161505451-161505451 on Build GRCh38
rs917352342 Chr.1: 161505370-161505370 on Build GRCh38
rs999815587 Chr.1: 161505617-161505617 on Build GRCh38
rs112591151 Chr.1: 161505546-161505546 on Build GRCh38
rs72717028 Chr.1: 161505324-161505324 on Build GRCh38
rs140132884 Chr.1: 161505695-161505695 on Build GRCh38
rs755997670 Chr.1: 161505545-161505545 on Build GRCh38
rs953835545 Chr.1: 161505343-161505343 on Build GRCh38
rs905323661 Chr.1: 161505267-161505267 on Build GRCh38
rs939952515 Chr.1: 161505381-161505381 on Build GRCh38
rs759636337 Chr.1: 161505534-161505534 on Build GRCh38
rs756503520 Chr.1: 161505293-161505293 on Build GRCh38
rs921027124 Chr.1: 161505342-161505342 on Build GRCh38
rs557302212 Chr.1: 161505507-161505507 on Build GRCh38
rs953805000 Chr.1: 161505676-161505676 on Build GRCh38
rs567320431 Chr.1: 161505656-161505656 on Build GRCh38
rs115266897 Chr.1: 161505383-161505383 on Build GRCh38
rs541104598 Chr.1: 161505310-161505310 on Build GRCh38
rs967269162 Chr.1: 161505309-161505309 on Build GRCh38
rs745350800 Chr.1: 161505506-161505506 on Build GRCh38
rs578116538 Chr.1: 161505351-161505351 on Build GRCh38
rs748659003 Chr.1: 161505517-161505517 on Build GRCh38
rs971893092 Chr.1: 161505657-161505657 on Build GRCh38
rs112207154 Chr.1: 161505736-161505736 on Build GRCh38
rs770401361 Chr.1: 161505521-161505521 on Build GRCh38
rs757115049 Chr.1: 161505557-161505557 on Build GRCh38

View Full Product Details