Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLOD5
Entrez Gene ID
392465
Gene Name
glyoxalase domain containing 5
Gene Aliases
-
Chromosome Location
Chr.X: 48761572 - 48773648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080489.2 NP_001073958.2 1 - 2 106 102

View Full Product Details