The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000206.2 | NP_000197.1 | 1 - 2 | 204 | 63 |
GenBank mRNA | AK298656.1 | - | 1 - 2 | 179 | 63 |
AK314932.1 | - | 1 - 2 | 148 | 63 | |
BC014972.1 | - | 1 - 2 | 116 | 63 | |
D11086.1 | - | 1 - 2 | 126 | 63 | |
DC345219.1 | - | 1 - 2 | 206 | 63 |