Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23563237-23563814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
UniGene
Hs.343244
Gene Chromosome Location
Chr.14: 23559563-23569056 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021743.1 XM_017021744.1 XM_017021745.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Gene Symbol
LOC102724814
Entrez Gene ID
102724814
Gene Name
uncharacterized LOC102724814
Gene Aliases
-
UniGene
Hs.569375
Gene Chromosome Location
Chr.14: 23561097-23568076 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021743.1 XM_017021744.1 XM_017021745.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601532 c.1346C>G p.A449G Chr.14: 23563444-23563444 on Build GRCh38
954810 c.1211G>A p.G404D Chr.14: 23563737-23563737 on Build GRCh38
954811 c.1153G>T p.E385* Chr.14: 23563795-23563795 on Build GRCh38
3987591 c.798-1G>A p.? Chr.14: 23563639-23563639 on Build GRCh38
6051091 c.1217T>C p.L406P Chr.14: 23563731-23563731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016637292 Chr.14: 23563278-23563278 on Build GRCh38
rs776340168 Chr.14: 23563568-23563568 on Build GRCh38
rs778617867 Chr.14: 23563509-23563509 on Build GRCh38
rs140628752 Chr.14: 23563451-23563451 on Build GRCh38
rs905811644 Chr.14: 23563264-23563264 on Build GRCh38
rs891645481 Chr.14: 23563351-23563351 on Build GRCh38
rs140030505 Chr.14: 23563415-23563415 on Build GRCh38
rs190256861 Chr.14: 23563353-23563353 on Build GRCh38
rs996120452 Chr.14: 23563651-23563651 on Build GRCh38
rs768435957 Chr.14: 23563694-23563694 on Build GRCh38
rs1000021465 Chr.14: 23563336-23563336 on Build GRCh38
rs377233339 Chr.14: 23563518-23563518 on Build GRCh38
rs898175702 Chr.14: 23563285-23563285 on Build GRCh38
rs866468608 Chr.14: 23563316-23563316 on Build GRCh38
rs754721186 Chr.14: 23563445-23563445 on Build GRCh38
rs761440759 Chr.14: 23563718-23563718 on Build GRCh38
rs757201793 Chr.14: 23563407-23563407 on Build GRCh38
rs756206729 Chr.14: 23563793-23563793 on Build GRCh38
rs996172603 Chr.14: 23563369-23563369 on Build GRCh38
rs745845243 Chr.14: 23563656-23563656 on Build GRCh38
rs558439142 Chr.14: 23563468-23563468 on Build GRCh38
rs996262752 Chr.14: 23563757-23563757 on Build GRCh38
rs200923646 Chr.14: 23563486-23563486 on Build GRCh38
rs184979172 Chr.14: 23563641-23563641 on Build GRCh38
rs543928791 Chr.14: 23563487-23563487 on Build GRCh38
rs574899141 Chr.14: 23563584-23563584 on Build GRCh38
rs201144415 Chr.14: 23563596-23563596 on Build GRCh38
rs372112838 Chr.14: 23563634-23563634 on Build GRCh38
rs770362556 Chr.14: 23563714-23563714 on Build GRCh38
rs748453641 Chr.14: 23563702-23563702 on Build GRCh38
rs749982998 Chr.14: 23563444-23563445 on Build GRCh38
rs1044335793 Chr.14: 23563313-23563313 on Build GRCh38
rs781231680 Chr.14: 23563640-23563640 on Build GRCh38
rs746085354 Chr.14: 23563380-23563380 on Build GRCh38
rs556896475 Chr.14: 23563708-23563708 on Build GRCh38
rs751773168 Chr.14: 23563622-23563622 on Build GRCh38
rs776215520 Chr.14: 23563695-23563695 on Build GRCh38
rs754208169 Chr.14: 23563365-23563365 on Build GRCh38
rs751542196 Chr.14: 23563488-23563488 on Build GRCh38
rs1001432768 Chr.14: 23563269-23563269 on Build GRCh38
rs757678536 Chr.14: 23563368-23563368 on Build GRCh38
rs1028921975 Chr.14: 23563665-23563665 on Build GRCh38
rs376640065 Chr.14: 23563462-23563462 on Build GRCh38
rs767727461 Chr.14: 23563777-23563777 on Build GRCh38
rs765684529 Chr.14: 23563482-23563482 on Build GRCh38
rs148842762 Chr.14: 23563481-23563481 on Build GRCh38
rs770192694 Chr.14: 23563722-23563722 on Build GRCh38
rs11626216 Chr.14: 23563668-23563668 on Build GRCh38
rs752351764 Chr.14: 23563501-23563501 on Build GRCh38
rs757801608 Chr.14: 23563445-23563445 on Build GRCh38
rs867073333 Chr.14: 23563443-23563444 on Build GRCh38
rs201169755 Chr.14: 23563604-23563604 on Build GRCh38
rs916990842 Chr.14: 23563785-23563785 on Build GRCh38
rs770909327 Chr.14: 23563505-23563505 on Build GRCh38
rs201586390 Chr.14: 23563417-23563417 on Build GRCh38
rs373872679 Chr.14: 23563500-23563500 on Build GRCh38
rs150451648 Chr.14: 23563453-23563453 on Build GRCh38
rs758704781 Chr.14: 23563388-23563388 on Build GRCh38
rs752305932 Chr.14: 23563343-23563343 on Build GRCh38
rs564344933 Chr.14: 23563775-23563775 on Build GRCh38
rs866292267 Chr.14: 23563555-23563555 on Build GRCh38
rs746634190 Chr.14: 23563564-23563567 on Build GRCh38
rs1015183032 Chr.14: 23563678-23563678 on Build GRCh38
rs111467384 Chr.14: 23563649-23563649 on Build GRCh38
rs145360826 Chr.14: 23563466-23563466 on Build GRCh38
rs1010699127 Chr.14: 23563352-23563352 on Build GRCh38
rs774194320 Chr.14: 23563430-23563430 on Build GRCh38
rs547285250 Chr.14: 23563675-23563675 on Build GRCh38
rs1009519823 Chr.14: 23563338-23563338 on Build GRCh38
rs754369597 Chr.14: 23563470-23563470 on Build GRCh38
rs375198511 Chr.14: 23563756-23563756 on Build GRCh38
rs771582263 Chr.14: 23563526-23563526 on Build GRCh38
rs949705488 Chr.14: 23563309-23563309 on Build GRCh38
rs760289303 Chr.14: 23563545-23563545 on Build GRCh38
rs781697725 Chr.14: 23563670-23563670 on Build GRCh38
rs780436116 Chr.14: 23563788-23563788 on Build GRCh38
rs775189531 Chr.14: 23563528-23563528 on Build GRCh38
rs373713459 Chr.14: 23563370-23563370 on Build GRCh38
rs752731458 Chr.14: 23563789-23563789 on Build GRCh38
rs1054381923 Chr.14: 23563547-23563547 on Build GRCh38
rs779942226 Chr.14: 23563391-23563391 on Build GRCh38
rs561657805 Chr.14: 23563666-23563666 on Build GRCh38
rs747117877 Chr.14: 23563414-23563414 on Build GRCh38
rs368289295 Chr.14: 23563683-23563683 on Build GRCh38
rs374332872 Chr.14: 23563335-23563335 on Build GRCh38
rs370083184 Chr.14: 23563585-23563585 on Build GRCh38
rs759093157 Chr.14: 23563446-23563446 on Build GRCh38
rs146957849 Chr.14: 23563613-23563613 on Build GRCh38
rs755447663 Chr.14: 23563582-23563597 on Build GRCh38
rs926994757 Chr.14: 23563506-23563506 on Build GRCh38
rs773744591 Chr.14: 23563740-23563740 on Build GRCh38
rs763408918 Chr.14: 23563755-23563755 on Build GRCh38
rs765019715 Chr.14: 23563422-23563423 on Build GRCh38
rs757116190 Chr.14: 23563644-23563644 on Build GRCh38
rs146876582 Chr.14: 23563428-23563428 on Build GRCh38
rs779809117 Chr.14: 23563452-23563452 on Build GRCh38
rs143631339 Chr.14: 23563418-23563418 on Build GRCh38
rs750830947 Chr.14: 23563484-23563484 on Build GRCh38
rs115840637 Chr.14: 23563337-23563337 on Build GRCh38
rs527534921 Chr.14: 23563674-23563674 on Build GRCh38
rs572740402 Chr.14: 23563444-23563444 on Build GRCh38
rs764695386 Chr.14: 23563362-23563362 on Build GRCh38
rs768282221 Chr.14: 23563551-23563551 on Build GRCh38
rs966010510 Chr.14: 23563364-23563364 on Build GRCh38
rs774636208 Chr.14: 23563767-23563767 on Build GRCh38
rs372502109 Chr.14: 23563684-23563684 on Build GRCh38

View Full Product Details