The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138383.2 | NP_612392.1 | 8 - 9 | 884 | 66 |
XM_005256236.4 | XP_005256293.2 | 7 - 8 | 1031 | 66 | |
XM_011523450.2 | XP_011521752.1 | 9 - 10 | 788 | 66 | |
XM_011523453.2 | XP_011521755.2 | 8 - 9 | 1040 | 66 | |
XM_011523454.2 | XP_011521756.2 | 7 - 8 | 1031 | 66 | |
XM_011523458.2 | XP_011521760.1 | 2 - 3 | 265 | 66 | |
XM_017023879.1 | XP_016879368.1 | 8 - 9 | 1040 | 66 | |
XM_017023880.1 | XP_016879369.1 | 8 - 9 | 1040 | 66 | |
XM_017023881.1 | XP_016879370.1 | 8 - 9 | 1040 | 66 | |
XM_017023882.1 | XP_016879371.1 | 8 - 9 | 1040 | 66 | |
XM_017023883.1 | XP_016879372.1 | 8 - 9 | 1040 | 66 | |
XM_017023884.1 | XP_016879373.1 | 8 - 9 | 1170 | 66 | |
GenBank mRNA | AB115770.1 | - | 8 - 9 | 624 | 66 |
BC002770.1 | - | 4 - 5 | 380 | 66 |