The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_038344.1 | 3 - 4 | 555 | 67 | |
NR_038345.1 | 2 - 3 | 383 | 67 | ||
GenBank mRNA | AB371456.1 | - | 2 - 3 | 229 | 67 |
BI521274.1 | - | 3 - 4 | 556 | 67 | |
CN311982.1 | - | 3 - 4 | 522 | 67 |