Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50908525-50909067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
UniGene
Hs.413366
Gene Chromosome Location
Chr.15: 50908569-51005900 on Build GRCh38
Transcripts
NM_001252127.1 NM_007347.4 XM_005254264.3 XM_006720447.3 XM_017022042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077294 c.104G>T p.R35M Chr.15: 50908882-50908882 on Build GRCh38
2216796 c.98C>T p.S33F Chr.15: 50908876-50908876 on Build GRCh38
5740342 c.1A>G p.M1V Chr.15: 50908779-50908779 on Build GRCh38
6538297 c.145A>G p.K49E Chr.15: 50908923-50908923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533962357 Chr.15: 50908758-50908758 on Build GRCh38
rs746511128 Chr.15: 50908891-50908891 on Build GRCh38
rs772294763 Chr.15: 50908741-50908742 on Build GRCh38
rs779273269 Chr.15: 50908974-50908974 on Build GRCh38
rs766330413 Chr.15: 50908843-50908843 on Build GRCh38
rs970696566 Chr.15: 50908564-50908564 on Build GRCh38
rs575583199 Chr.15: 50909031-50909031 on Build GRCh38
rs781171572 Chr.15: 50908789-50908789 on Build GRCh38
rs1035869608 Chr.15: 50908723-50908723 on Build GRCh38
rs868781587 Chr.15: 50908548-50908548 on Build GRCh38
rs981669378 Chr.15: 50908572-50908572 on Build GRCh38
rs946447075 Chr.15: 50908816-50908816 on Build GRCh38
rs1052143968 Chr.15: 50908589-50908589 on Build GRCh38
rs780539825 Chr.15: 50908878-50908878 on Build GRCh38
rs370276551 Chr.15: 50908818-50908818 on Build GRCh38
rs775028915 Chr.15: 50908810-50908810 on Build GRCh38
rs995994792 Chr.15: 50908973-50908973 on Build GRCh38
rs760293485 Chr.15: 50908759-50908759 on Build GRCh38
rs745897561 Chr.15: 50908742-50908759 on Build GRCh38
rs765289252 Chr.15: 50908840-50908840 on Build GRCh38
rs748241372 Chr.15: 50908790-50908790 on Build GRCh38
rs766231443 Chr.15: 50908926-50908926 on Build GRCh38
rs199970888 Chr.15: 50908946-50908946 on Build GRCh38
rs779466797 Chr.15: 50908871-50908871 on Build GRCh38
rs868114700 Chr.15: 50908760-50908777 on Build GRCh38
rs2306329 Chr.15: 50908642-50908642 on Build GRCh38
rs777156091 Chr.15: 50908677-50908677 on Build GRCh38
rs778137187 Chr.15: 50908806-50908806 on Build GRCh38
rs774920356 Chr.15: 50908822-50908822 on Build GRCh38
rs566703744 Chr.15: 50908744-50908744 on Build GRCh38
rs771632650 Chr.15: 50908738-50908738 on Build GRCh38
rs762804786 Chr.15: 50908916-50908916 on Build GRCh38
rs987333956 Chr.15: 50908884-50908884 on Build GRCh38
rs920925586 Chr.15: 50908834-50908834 on Build GRCh38
rs762851995 Chr.15: 50908842-50908842 on Build GRCh38
rs1057518523 Chr.15: 50908776-50908776 on Build GRCh38
rs553522152 Chr.15: 50908807-50908807 on Build GRCh38
rs1052138905 Chr.15: 50908709-50908709 on Build GRCh38
rs995543214 Chr.15: 50908707-50908707 on Build GRCh38
rs139243461 Chr.15: 50908593-50908593 on Build GRCh38
rs977179256 Chr.15: 50908860-50908860 on Build GRCh38
rs2306330 Chr.15: 50908702-50908702 on Build GRCh38
rs760224898 Chr.15: 50908823-50908823 on Build GRCh38
rs916182807 Chr.15: 50908561-50908561 on Build GRCh38
rs763655101 Chr.15: 50908824-50908824 on Build GRCh38
rs143027953 Chr.15: 50908820-50908820 on Build GRCh38
rs750754501 Chr.15: 50908975-50908975 on Build GRCh38
rs1023011674 Chr.15: 50908981-50908981 on Build GRCh38
rs761496023 Chr.15: 50908908-50908908 on Build GRCh38
rs1033919223 Chr.15: 50908646-50908646 on Build GRCh38
rs550277890 Chr.15: 50908701-50908701 on Build GRCh38
rs889025322 Chr.15: 50908712-50908712 on Build GRCh38
rs778157257 Chr.15: 50908853-50908853 on Build GRCh38
rs890964725 Chr.15: 50908693-50908693 on Build GRCh38
rs76674534 Chr.15: 50908961-50908961 on Build GRCh38
rs374083032 Chr.15: 50908739-50908739 on Build GRCh38
rs1026493869 Chr.15: 50908614-50908614 on Build GRCh38
rs755119255 Chr.15: 50908788-50908788 on Build GRCh38
rs946406375 Chr.15: 50908591-50908591 on Build GRCh38
rs773930982 Chr.15: 50908560-50908560 on Build GRCh38
rs937871666 Chr.15: 50908751-50908765 on Build GRCh38
rs899130633 Chr.15: 50908735-50908735 on Build GRCh38
rs962712252 Chr.15: 50908559-50908559 on Build GRCh38
rs771366562 Chr.15: 50908808-50908808 on Build GRCh38
rs758727248 Chr.15: 50908979-50908979 on Build GRCh38
rs928805961 Chr.15: 50908581-50908581 on Build GRCh38
rs982103605 Chr.15: 50908720-50908720 on Build GRCh38
rs748951930 Chr.15: 50908549-50908549 on Build GRCh38
rs1002848255 Chr.15: 50909037-50909037 on Build GRCh38
rs780257873 Chr.15: 50908747-50908748 on Build GRCh38
rs557031232 Chr.15: 50909024-50909024 on Build GRCh38
rs749657602 Chr.15: 50908737-50908737 on Build GRCh38
rs867144603 Chr.15: 50908772-50908772 on Build GRCh38
rs747259387 Chr.15: 50908758-50908761 on Build GRCh38
rs564376715 Chr.15: 50908586-50908586 on Build GRCh38
rs546316822 Chr.15: 50908613-50908613 on Build GRCh38
rs913626882 Chr.15: 50908590-50908590 on Build GRCh38
rs1023372999 Chr.15: 50908713-50908713 on Build GRCh38
rs367555487 Chr.15: 50908955-50908955 on Build GRCh38
rs756975040 Chr.15: 50908774-50908774 on Build GRCh38
rs756349659 Chr.15: 50908804-50908804 on Build GRCh38
rs751457873 Chr.15: 50908938-50908938 on Build GRCh38
rs776206463 Chr.15: 50908835-50908835 on Build GRCh38
rs754065099 Chr.15: 50908865-50908865 on Build GRCh38
rs750308874 Chr.15: 50908778-50908778 on Build GRCh38
rs757519956 Chr.15: 50908868-50908868 on Build GRCh38
rs924365071 Chr.15: 50908869-50908869 on Build GRCh38
rs898558625 Chr.15: 50908705-50908705 on Build GRCh38
rs768160491 Chr.15: 50908902-50908902 on Build GRCh38
rs762870498 Chr.15: 50908907-50908907 on Build GRCh38
rs373684336 Chr.15: 50908845-50908845 on Build GRCh38
rs970375919 Chr.15: 50908715-50908715 on Build GRCh38
rs377511016 Chr.15: 50908728-50908728 on Build GRCh38
rs375931068 Chr.15: 50908775-50908775 on Build GRCh38
rs112414735 Chr.15: 50908766-50908766 on Build GRCh38
rs895281980 Chr.15: 50909030-50909030 on Build GRCh38
rs151135245 Chr.15: 50908911-50908911 on Build GRCh38
rs750190505 Chr.15: 50908841-50908841 on Build GRCh38
rs760693526 Chr.15: 50908943-50908943 on Build GRCh38
rs991130873 Chr.15: 50908687-50908687 on Build GRCh38
rs1004015993 Chr.15: 50908991-50908991 on Build GRCh38
rs763788543 Chr.15: 50908764-50908764 on Build GRCh38
rs879834435 Chr.15: 50908573-50908573 on Build GRCh38
rs772519865 Chr.15: 50908877-50908877 on Build GRCh38
rs766895302 Chr.15: 50908756-50908756 on Build GRCh38
rs939853762 Chr.15: 50908889-50908889 on Build GRCh38
rs1016385590 Chr.15: 50908592-50908592 on Build GRCh38
rs906633556 Chr.15: 50908988-50908988 on Build GRCh38
rs568046524 Chr.15: 50908641-50908641 on Build GRCh38
rs773502598 Chr.15: 50908732-50908732 on Build GRCh38
rs759132322 Chr.15: 50908753-50908753 on Build GRCh38
rs770031390 Chr.15: 50908729-50908729 on Build GRCh38
rs772608764 Chr.15: 50908821-50908821 on Build GRCh38
rs756152684 Chr.15: 50908852-50908852 on Build GRCh38
rs746230196 Chr.15: 50908876-50908876 on Build GRCh38
rs12908516 Chr.15: 50908557-50908557 on Build GRCh38
rs539784751 Chr.15: 50908736-50908736 on Build GRCh38
rs958647234 Chr.15: 50908672-50908672 on Build GRCh38
rs989650049 Chr.15: 50908812-50908813 on Build GRCh38

View Full Product Details