The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136105.2 | NP_001129577.1 | 4 - 5 | 854 | 70 |
NM_001271457.1 | NP_001258386.1 | 4 - 5 | 599 | 70 | |
NM_001271458.1 | NP_001258387.1 | 4 - 5 | 423 | 70 | |
XM_017017196.1 | XP_016872685.1 | 5 - 6 | 503 | 70 | |
XM_017017197.1 | XP_016872686.1 | 3 - 4 | 1594 | 70 | |
XM_017017198.1 | XP_016872687.1 | 4 - 5 | 459 | 70 | |
XM_017017199.1 | XP_016872688.1 | 4 - 5 | 622 | 70 | |
XM_017017200.1 | XP_016872689.1 | 4 - 5 | 907 | 70 | |
GenBank mRNA | AK292195.1 | - | 4 - 5 | 854 | 70 |
BC042557.1 | - | 4 - 5 | 665 | 70 | |
BC110079.1 | - | 3 - 4 | 322 | 70 | |
BQ893717.1 | - | - | 629 | 70 | |
HY114242.1 | - | 4 - 5 | 424 | 70 | |
KF740510.1 | - | 4 - 5 | 799 | 70 | |
KF740511.1 | - | 4 - 5 | 414 | 70 | |
KF740512.1 | - | 5 - 6 | 519 | 70 | |
KF740513.1 | - | 5 - 6 | 645 | 70 | |
KF740514.1 | - | 5 - 6 | 997 | 70 |