Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11orf94
Entrez Gene ID
143678
Gene Name
chromosome 11 open reading frame 94
Gene Aliases
-
Chromosome Location
Chr.11: 45906534 - 45907282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080446.2 NP_001073915.2 1 - 2 135 86

View Full Product Details