Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46662076-46662610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2920750 c.1969G>A p.A657T Chr.16: 46662341-46662341 on Build GRCh38
4185773 c.1940A>G p.E647G Chr.16: 46662370-46662370 on Build GRCh38
2920751 c.1859A>T p.D620V Chr.16: 46662451-46662451 on Build GRCh38
970759 c.2036C>T p.S679F Chr.16: 46662274-46662274 on Build GRCh38
5462552 c.1828-9G>C p.? Chr.16: 46662491-46662491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890947445 Chr.16: 46662575-46662575 on Build GRCh38
rs751044238 Chr.16: 46662167-46662167 on Build GRCh38
rs375643041 Chr.16: 46662288-46662288 on Build GRCh38
rs371077269 Chr.16: 46662234-46662234 on Build GRCh38
rs769692699 Chr.16: 46662354-46662354 on Build GRCh38
rs868074638 Chr.16: 46662219-46662219 on Build GRCh38
rs368691901 Chr.16: 46662200-46662200 on Build GRCh38
rs748012697 Chr.16: 46662339-46662339 on Build GRCh38
rs748025800 Chr.16: 46662500-46662500 on Build GRCh38
rs761462194 Chr.16: 46662215-46662215 on Build GRCh38
rs941466302 Chr.16: 46662222-46662222 on Build GRCh38
rs559637051 Chr.16: 46662336-46662336 on Build GRCh38
rs775461699 Chr.16: 46662171-46662171 on Build GRCh38
rs776389848 Chr.16: 46662334-46662334 on Build GRCh38
rs111445265 Chr.16: 46662134-46662134 on Build GRCh38
rs776495649 Chr.16: 46662213-46662213 on Build GRCh38
rs765978232 Chr.16: 46662247-46662247 on Build GRCh38
rs138794859 Chr.16: 46662429-46662429 on Build GRCh38
rs768410261 Chr.16: 46662211-46662211 on Build GRCh38
rs756550058 Chr.16: 46662415-46662415 on Build GRCh38
rs567148854 Chr.16: 46662564-46662564 on Build GRCh38
rs569735490 Chr.16: 46662405-46662405 on Build GRCh38
rs201998902 Chr.16: 46662433-46662433 on Build GRCh38
rs188286943 Chr.16: 46662452-46662452 on Build GRCh38
rs764451293 Chr.16: 46662252-46662252 on Build GRCh38
rs376700700 Chr.16: 46662303-46662303 on Build GRCh38
rs188831675 Chr.16: 46662249-46662249 on Build GRCh38
rs764695026 Chr.16: 46662416-46662416 on Build GRCh38
rs900037539 Chr.16: 46662320-46662320 on Build GRCh38
rs754253874 Chr.16: 46662256-46662256 on Build GRCh38
rs746999621 Chr.16: 46662294-46662294 on Build GRCh38
rs780197148 Chr.16: 46662486-46662486 on Build GRCh38
rs1009202678 Chr.16: 46662406-46662406 on Build GRCh38
rs182929141 Chr.16: 46662240-46662240 on Build GRCh38
rs377476545 Chr.16: 46662229-46662229 on Build GRCh38
rs765025576 Chr.16: 46662179-46662179 on Build GRCh38
rs373245295 Chr.16: 46662220-46662220 on Build GRCh38
rs780111914 Chr.16: 46662290-46662290 on Build GRCh38
rs1038002002 Chr.16: 46662443-46662443 on Build GRCh38
rs917231636 Chr.16: 46662107-46662107 on Build GRCh38
rs145406665 Chr.16: 46662365-46662365 on Build GRCh38
rs168745 Chr.16: 46662372-46662372 on Build GRCh38
rs547369855 Chr.16: 46662275-46662275 on Build GRCh38
rs758632241 Chr.16: 46662453-46662453 on Build GRCh38
rs758539394 Chr.16: 46662276-46662276 on Build GRCh38
rs765010319 Chr.16: 46662227-46662227 on Build GRCh38
rs779153929 Chr.16: 46662263-46662263 on Build GRCh38
rs373895312 Chr.16: 46662527-46662527 on Build GRCh38
rs758922774 Chr.16: 46662409-46662409 on Build GRCh38
rs769781944 Chr.16: 46662526-46662526 on Build GRCh38
rs757674131 Chr.16: 46662427-46662427 on Build GRCh38
rs757523799 Chr.16: 46662261-46662261 on Build GRCh38
rs549129612 Chr.16: 46662487-46662487 on Build GRCh38
rs144997996 Chr.16: 46662398-46662398 on Build GRCh38
rs754978958 Chr.16: 46662492-46662492 on Build GRCh38
rs781241576 Chr.16: 46662497-46662497 on Build GRCh38
rs754270736 Chr.16: 46662419-46662419 on Build GRCh38

View Full Product Details