Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRPRB
Entrez Gene ID
58477
Gene Name
SRP receptor beta subunit
Gene Aliases
APMCF1
Chromosome Location
Chr.3: 133784033 - 133821492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021203.3 NP_067026.3 4 - 5 605 88
GenBank mRNA AF086914.1 - 3 - 4 343 88
AF141882.3 - 3 - 4 336 88
AK027525.1 - 3 - 4 336 88
AK075531.1 - 3 - 4 374 88
BC063001.1 - 3 - 4 348 88
BC065299.1 - 3 - 4 320 88

View Full Product Details